Skip to Content
Merck

Skip To

31394

Dextran

enzymatic synth.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
[C6H10O5]n
CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
EC Number:
232-677-5
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

mol wt

Mr ~1,500

color

colorless to almost colorless

solubility

water: 1.5 g/10 mL, clear

storage temp.

room temp

SMILES string

O1C(C(C(C(C1CO)O)O)O)OCC2OC(C(C(C2O)O)O)OCC(O)C(O)C(O)C(O)C=O

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Application

Dextran is a branched glucan composed of linear α(1→6) linked glucose units and α (1→3) link initiated branches. Dextran ranges in size from 10,000 to 150,000 Kd. Dextrans are used in many applications as volume extenders, stabilizers, matrix components, binding platforms, lubricants and physical structure components.

Other Notes

To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
314300026800271
solubility

water: 1.5 g/10 mL, clear

solubility

-

solubility

-

solubility

-

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100

form

powder

form

-

form

powder or crystals

form

powder or crystals

storage temp.

room temp

storage temp.

-

storage temp.

-

storage temp.

-

color

colorless to almost colorless

color

-

color

-

color

-

mol wt

Mr ~1,500

mol wt

Mp 1,000-400,000

mol wt

-

mol wt

-


Still not finding the right product?

Explore all of our products under Dextran


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
31394-100G04061833030790
31394-25G04061833030806
31394-5G04061833030813

Questions

Reviews

No rating value

Active Filters