Skip to Content
Merck

Skip To

BOD2000

BOD 2000 mg/L Calibration Standard

certified reference material

Synonym(s):

Biochemical Oxygen Demand Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

agency

suitable for SM 5210

CofA

current certificate can be downloaded

manufacturer/tradename

RTC BOD2000-500

concentration

in H2O

application(s)

environmental

format

matrix material

storage temp.

2-8°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

This CRM is intended for the calibration and validation of analytical instruments used to measure biological oxygen demand (BOD).

Features and Benefits

  • Verified and qualified in an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to the SI and higher order standards from NIST
  • Assigned value based on a purity determination by mass balance and gravimetrically prepared value
  • Homogeneity assessed in accordance with ISO Guide 35
  • Available in 500 mL high-density polyethylene bottle for an assured safe delivery
  • Accompanied with a certificate of analysis (CoA) having information on the expiry date for an effective audit
  • Prepared using deionized water as a solvent

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BOD200BOD1000BOD500
application(s)

environmental

application(s)

environmental

application(s)

environmental

application(s)

environmental

format

matrix material

format

matrix material

format

matrix material

format

matrix material

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

agency

suitable for SM 5210

agency

suitable for SM 5210

agency

suitable for SM 5210

agency

suitable for SM 5210

CofA

current certificate can be downloaded

CofA

certificate is enclosed in each package

CofA

current certificate can be downloaded

CofA

current certificate can be downloaded

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300


wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters