Skip to Content
Merck

Skip To

97151

Hederacoside C

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C59H96O26
CAS Number:
Molecular Weight:
1221.38
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
EC Number:
238-072-2
Beilstein/REAXYS Number:
4229541
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

mp

222 °C (dec.) (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

[H][C@]12CC=C3C4CC(C)(C)CC[C@@]4(CC[C@@]3(C)[C@]1(C)CCC5[C@](C)(CO)[C@H](CC[C@]25C)O[C@@H]6OC[C@H](O)[C@H](O)[C@H]6O[C@@H]7O[C@@H](C)[C@H](O)[C@@H](O)[C@H]7O)C(=O)O[C@@H]8O[C@H](CO[C@@H]9O[C@H](CO)[C@@H](O[C@@H]%10O[C@@H](C)[C@H](O)[C@@H](O)[C@H]%10O)[C@H](O)[C@H]9O)[C@@H](O)[C@H](O)[C@H]8O

InChI

1S/C59H96O26/c1-24-34(63)38(67)42(71)49(78-24)83-46-29(20-60)80-48(45(74)41(46)70)77-22-30-37(66)40(69)44(73)51(81-30)85-53(75)59-17-15-54(3,4)19-27(59)26-9-10-32-55(5)13-12-33(56(6,23-61)31(55)11-14-58(32,8)57(26,7)16-18-59)82-52-47(36(65)28(62)21-76-52)84-50-43(72)39(68)35(64)25(2)79-50/h9,24-25,27-52,60-74H,10-23H2,1-8H3/t24-,25-,27?,28-,29+,30+,31?,32+,33-,34-,35-,36-,37+,38+,39+,40-,41+,42+,43+,44+,45+,46+,47+,48+,49-,50-,51-,52-,55-,56-,57+,58+,59-/m0/s1

InChI key

RYHDIBJJJRNDSX-WBOYGZCJSA-N

General description

Hederacoside C is a natural triterpenoid saponin. It is the primary bioactive ingredient of traditional Chinese herbal medicines, isolated from Hedera helix, also known as common ivy or English ivy. It is known for its expectorant, bronchodilator, antibacterial, and bronchospasmolytic properties in the treatment of respiratory disorders.

Application

This analytical standard can also be used as follows:
  • Development of a bioanalytical method to measure hederacoside c in rat plasma samples by ultra high-performance liquid chromatographic-tandem mass spectrometric (UPLC-MS/MS) method
  • Simultaneous analysis of six natural compounds isolated from ivy leaf extract samples by UHPLC coupled with electrospray ionization-triple quadrupole mass spectrometeric (ESI-MS/MS) detection
  • Ultra-high-performance liquid chromatography-quadrupole time-of-flight mass spectrometry (UHPLC‐Q‐TOF‐MS/MS) based phytochemical profiling of Hedera helix for pharmaceutical applications

Packaging

Bottomless glass bottle. The content inside is inserted in a fused cone.

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHL8922100930585PHL84218
application(s)

food and beverages

application(s)

-

application(s)

-

application(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

-

technique(s)

-

format

neat

format

-

format

-

format

-

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

-

grade

analytical standard

grade

primary reference standard

grade

primary reference standard

grade

primary reference standard

storage temp.

2-8°C

storage temp.

-

storage temp.

-

storage temp.

-


Still not finding the right product?

Explore all of our products under Hederacoside C


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



A validated RP HPLC-PAD method for the determination of hederacoside C in Ivy-Thyme cough syrup
Khdair A, et al.
International Journal of Analytical Chemistry (2010)
S H Cho et al.
Archives of pharmacal research, 14(1), 19-24 (1991-03-01)
One new triterpenoidal saponin, saponin F(2) has been isolated from the bark of Kalopanax pictum Nakai var. typicum (Araliaceae), together with one known saponin, kizuta saponin K12 (1). On the basis of chemico-spectral evidences, the structure of 2 has been
Eun-Ha Joh et al.
Phytotherapy research : PTR, 26(4), 546-551 (2011-09-20)
The stem-bark of Kalopanax pictus (KP, family Araliaceae), which contains triterpenoid saponins, has been shown to exhibit anticarcinogenic, antiinflammatory, antirheumatoid and antidiabetic activities. In a preliminary study, a KP methanol extract demonstrated acetylcholinesterase activity in vitro and memory enhancement in



Global Trade Item Number

SKUGTIN
97151-50MG-F04061835560127

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters