The nominal concentrations can be found in the contents subsection of the description section on this product webpage. The actual concentrations can be found in the documents section, by selecting the individual lot of interest.
Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
analytical standard
Quality Level
quality
6 PCB according to TrinkwV (BRD) method
shelf life
limited shelf life, expiry date on the label
concentration
in isooctane
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
application(s)
environmental
format
multi-component solution
storage temp.
2-8°C
Application
Other Notes
2,4,4′-PCB, 2,2′,5,5′-PCB, 2,2′,4,5,5′-PCB, 2,2′,3,4,4′,5-PCB, 2,2′,4,4′,5,5′-PCB, 2,2′,3,4,4′,5,5′-PCB
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable |
| format multi-component solution | format multi-component solution | format neat | format neat |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| grade analytical standard | grade analytical standard | grade analytical standard | grade analytical standard |
| storage temp. 2-8°C | storage temp. - | storage temp. - | storage temp. - |
| concentration in isooctane | concentration 10 μg/mL in isooctane | concentration - | concentration - |
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Central nervous system
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
39.2 °F
flash_point_c
4 °C
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 36906-10ML | 04061837579653 |
| 36906-2ML | 04061831832952 |
-
Wo kann ich sehen welche Konzentration der Standard hat? Wo sehe ich welche Kongenere in den Verschiedenen "PCB-Mix" enthalten sind. Es fehlt mir die übersicht, welche früher durch den Katalog geboten wurde.
1 answer-
Helpful?
-







