Skip to Content
Merck

Skip To

TAQ-RO

Roche

Taq DNA Polymerase, 5 U/μl

optimum pH ~9.0 (20 °C), optimum reaction temp. 72 °C

Synonym(s):

dna amplification, pcr, polymerase, pcr, primer extension, dna amplification, primer extension

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
41106300
EC Number:
NACRES:
NA.54
Specific activity:
5 U/μL
Biological source:
bacterial (Thermus aquaticus)
Recombinant:
expressed in E. coli
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bacterial (Thermus aquaticus)

recombinant

expressed in E. coli

grade

Molecular Biology

form

liquid

usage

sufficient for ≤1,000 reactions (11146173001), sufficient for ≤10,000 reactions (11435094001), sufficient for ≤2,000 reactions (11418432001), sufficient for ≤5,000 reactions (11596594001), sufficient for ≤200 reactions (11146165001)

specific activity

5 U/μL

mol wt

95 kDa

packaging

pkg of 1,000 U (11418432001 [4 x 250 U]), pkg of 2,500 U (11596594001 [10 x 250 U]), pkg of 5,000 U (11435094001 [20 x 250 U]), pkg of 100 U (11146165001), pkg of 500 U (11146173001 [1 x 500 U])

manufacturer/tradename

Roche

parameter

72 °C optimum reaction temp.

technique(s)

DNA sequencing: suitable, PCR: suitable

color

colorless

optimum pH

~9.0 (20 °C)

solubility

water: miscible

suitability

suitable for molecular biology

NCBI accession no.

UniProt accession no.

application(s)

genomic analysis
life science and biopharma

foreign activity

Nicking activity 30 units, none detected, Ribonuclease 30 units, none detected, Unspecific endonucleases 30 units, none detected

storage temp.

−20°C

General description

Taq DNA Polymerase is a highly processive 5′→3′ DNA polymerase that lacks 3′→5′ exonuclease activity. It is a single polypeptide chain with a molecular weight of approximately 95 kDa.
Taq DNA Polymerase was originally isolated from the thermophilic eubacterium Thermus aquaticus BM, a strain lacking Taq I restriction endonuclease. The enzyme was cloned in E.coli.

Application

Taq DNA Polymerase can be used in simple, routine PCR. Roche Applied Science Taq DNA polymerase is held to rigorous purity and quality standards. This preparation of recombinant Taq DNA Polymerase can be applied for:
  • PCR
  • RT-PCR[1]
  • qPCR[2][3]
  • Other primer-extension reactions, such as sequencing and labeling

Features and Benefits

Reliable reproducible results:
High lot-to-lot consistency.
No need to test each lot:
Taq DNA Polymerase is rigorously tested.
Prevent PCR carryover:
dUTP incorporation combination with Uracil-DNA Glycosylase prevents PCR cross-contamination.

Packaging

1 kit containing 2 components

Analysis Note

Routine assays have medium size amplicons and 50% GC content. Taq DNA Polymerase has no proofreading or hot start features. It is optimally active at +75°C and pH 9.
Lack of restriction endonuclease:
The enzyme originally isolated from T. aquaticus BM lacks Taq I restriction endonuclease activity.Each lot is PCR tested using λDNA. Each lot is also tested for the absence of exo- and endonucleases, and nicking activities according to the current Quality Control procedures.

Other Notes

For life science research only. Not for use in diagnostic procedures.
One unit Taq DNA Polymerase is defined as the amount of enzyme that incorporates 10 nmol of total deoxyribonucleosidetriphosphates into acid precipitable DNA within 60 min at +65 °C under the assay conditions given above.

Unit Assay: Incubation buffer:
67 mM Tris/HCl; pH 8.3/25 °C, 5 mM MgCl2, 10 mM Mercaptoethanol, 0.2% Polydocanol, 0.2 mg/ml Gelatine, 0.2 mM each dATP, dGTP, dTTP and 0.1 mM dCTP.

Incubation procedure:
M13mp9ss, M13 primer (17mer) and 1 μCi (α-32P) dCTP are incubated with suitable dilutions of Taq DNA Polymerase in 50 μl incubation buffer at +65 °C for 60 minutes. The amount of incorporated dNTPs is determined by trichloroacetic acid precipitation.

Volume Activity: 5 U/μl

Legal Information

Use of this product is covered by US patent claims and corresponding patent claims outside the US. The purchase of this product includes a limited, non-transferable immunity from suit under the foregoing patent claims for using only this amount of product for the purchaser′s own internal research. No right under any other patent claim, no right to perform any patented method, and no right to perform commercial services of any kind, including without limitation reporting the results of purchaser′s activities for a fee or other commercial consideration, is conveyed expressly, by implication, or by estoppel. This product is for research use only. Diagnostic uses under Roche patent claims require a separate license from Roche. Further information on purchasing licenses may be obtained by contacting the Director of Licensing, Applied Biosystems, 850 Lincoln Centre Drive, Foster City, California 94404, USA.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
TAQL-ROTAQN-RO03734927001
grade

Molecular Biology

grade

-

grade

-

grade

-

biological source

bacterial (Thermus aquaticus)

biological source

bacterial (Thermus aquaticus BM)

biological source

bacterial (Thermus aquaticus BM)

biological source

-

specific activity

5 U/μL

specific activity

1 U/μL

specific activity

-

specific activity

≥13000 units/mg protein

technique(s)

DNA sequencing: suitable, PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

technique(s)

PCR: suitable

application(s)

genomic analysis
life science and biopharma

application(s)

genomic analysis
life science and biopharma

application(s)

genomic analysis
life science and biopharma

application(s)

-

suitability

suitable for molecular biology

suitability

suitable for PCR, suitable for molecular biology

suitability

suitable for PCR and automated sequencing reactions

suitability

-


Kit Components Only

Product No.
Description

  • Taq DNA Polymerase 5 U/μl

  • PCR Buffer with MgCl<sub>2</sub> 10x concentrated

  • MgCl<sub>2</sub> Stock Solution

  • PCR Buffer without MgCl<sub>2</sub>

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



H N Chinivasagam et al.
Journal of applied microbiology, 103(2), 418-426 (2007-07-26)
To investigate the occurrence and levels of Arcobacter spp. in pig effluent ponds and effluent-treated soil. A Most Probable Number (MPN) method was developed to assess the levels of Arcobacter spp. in seven pig effluent ponds and six effluent-treated soils
Mónica Alejandra Rosales-Reynoso et al.
Archives of medical research, 41(2), 110-118 (2010-05-18)
Although sporadic cases of cancer in patients with fragile X syndrome (FXS) have been reported, extensive studies carried out in Denmark and Finland concluded that cancer incidence in these patients is lower than in the general population. On the other
Pattabhiraman Shankaranarayanan et al.
Nature protocols, 7(2), 328-338 (2012-01-28)
Linear amplification of DNA (LinDA) by T7 polymerase is a versatile and robust method for generating sufficient amounts of DNA for genome-wide studies with minute amounts of cells. LinDA can be coupled to a great number of global profiling technologies.



Global Trade Item Number

SKUGTIN
1114616500104061838698995
1159659400104061837893476
1114617300104061837893414
1143509400104061837893438
1141843200104061837893421