Skip to Content
Merck

Skip To

SFUKB

Roche

KAPA SYBR® FAST

suitable for qPCR, 2 ×, Universal

Synonym(s):

SYBR, qPCR

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.55
UNSPSC Code:
41106300
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 100 reactions, sufficient for 1000 reactions, sufficient for 500 reactions, sufficient for 5000 reactions

shelf life

≤12 mo.

feature

dNTPs included, hotstart

packaging

kit of 1 mL (100 x 20 μL rxn; KK4600), kit of 10 mL (1000 x 20 μL rxn; KK4602), kit of 5 mL (500 x 20 μL rxn; KK4601), kit of 50 mL (5000 x 20 μL rxn; KK4618)

manufacturer/tradename

Roche

concentration

2 ×

technique(s)

qPCR: suitable

input

purified DNA

compatibility

for use with Agilent Mx3000P, for use with Agilent Mx3005P, for use with Agilent Mx4000, for use with Applied Biosystems® 5700, for use with Applied Biosystems® 7000, for use with Applied Biosystems® 7300, for use with Applied Biosystems® 7500, for use with Applied Biosystems® 7700, for use with Applied Biosystems® 7900HT, for use with Applied Biosystems® ViiA 7, for use with QuantStudio, for use with StepOne, for use with StepOnePlus

detection method

SYBR® Green

storage temp.

−20°C

General description

KAPA SYBR® FAST qPCR Kits contain the first DNA polymerase engineered via directed evolution to be more tolerant of SYBR Green I dye inhibition.

The improved robustness, processivity, and speed of KAPA SYBR FAST qPCR Kits result in consistently high amplification efficiencies enabling more accurate relative quantification for gene expression analysis. KAPA SYBR FAST qPCR Kits, developed to perform optimally in stringent real-time PCR reaction conditions, exhibit dramatic improvements to signal-to-noise ratio (fluorescence), quantification cycle (Cq), linearity, and sensitivity.

Application

KAPA SYBR® FAST has been used in:
  • Gene expression
  • Gene knockdown validation
  • Microarray validation
  • ChIP analysis
  • Low copy detection
  • Absolute quantification of NGS libraries
  • Quantitative real-time PCR

Features and Benefits

Quantitate changes in gene expression more accurately
  • High reaction efficiency between 95 - 105% improves accuracy and reproducibility.
  • Unbiased efficiency across a wide range of GC contents and amplicon lengths.

Detect low copy and difficult targets consistently
  • Improved processivity results in earlier Cq scores.
  • Higher fluorescence detection across varying AT- and GC-rich targets.
  • Novel enzyme is resistant to SYBR® inhibition.

Complete real-time PCR runs in just 40 minutes
  • 55% shorter run times with fast cycling protocol.
  • Maintain high performance when switching from standard to fast protocols.

Quick Notes:
  • This kit contains an engineered enzyme optimized for qPCR using SYBR Green I dye chemistry.
  • The 2X master mix contains a proprietary buffer. Together with the novel enzyme, this improves amplification efficiency of both GC- and AT-rich targets.
  • 20 sec initial denaturation at 95°C is sufficient for enzyme activation. When working with complex templates, an initial denaturation of 3 min is recommended.
  • For 3-step cycling, use 20 sec for primer annealing and 1 sec for extension/data acquisition at 72°C.
  • Do not exceed 25 μL reaction volumes.

Preparation Note

Always ensure that components have been fully thawed and thoroughly mixed before use. KAPA SYBR® FAST qPCR Master Mix (2X) may not freeze solidly, even when stored at -20°C.

The SYBR Green I dye contained in KAPA SYBR FAST qPCR Master Mix (2X) and ROX/fluorescein dyes (depending on kit configuration) are light sensitive. Exposure to direct light for an extended period of time will result in loss of fluorescent signal intensity.

KAPA SYBR FAST qPCR Master Mix (2X) is stable through 30 freeze-thaw cycles. Ensure that all reagents are stored protected from light at -20°C when not in use. When protected from light, reagents are stable in the dark at 4°C for at least one week and may be stored at this temperature for short-term use, provided that they do not become contaminated with microbes and/or nucleases.

Analysis Note

All kit components are subjected to stringent functional quality control, are free of detectable contaminating exo and endonuclease activity, and meet strict requirements with respect to DNA contamination.

Other Notes

For Research Use Only. Not for use in diagnostic procedures.

Legal Information

Applied Biosystems is a registered trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
QuantStudio is a trademark of IROA Technologies LLC
SYBR is a registered trademark of Thermo Fisher Scientific or its subsidiaries
StepOne is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
StepOnePlus is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SFABIKBSFLRABSFLCKB
technique(s)

qPCR: suitable

technique(s)

qPCR: suitable

technique(s)

qPCR: suitable

technique(s)

qPCR: suitable

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

compatibility

for use with Agilent Mx3000P, for use with Agilent Mx4000, for use with Applied Biosystems® 7000, for use with Applied Biosystems® 7500, for use with Applied Biosystems® 7900HT, for use with QuantStudio, for use with StepOnePlus, for use with Agilent Mx3005P, for use with Applied Biosystems® 7300, for use with Applied Biosystems® ViiA 7, for use with Applied Biosystems® 5700, for use with StepOne, for use with Applied Biosystems® 7700

compatibility

for use with Applied Biosystems® 5700, for use with Applied Biosystems® 7000, for use with Applied Biosystems® 7300, for use with Applied Biosystems® 7700, for use with Applied Biosystems® 7900HT, for use with StepOne, for use with StepOnePlus

compatibility

for use with Agilent Mx3000P, for use with Agilent Mx3005P, for use with Agilent Mx4000, for use with Applied Biosystems® 7500, for use with Applied Biosystems® ViiA 7, for use with QuantStudio

compatibility

for use with Bio-Rad CFX96, for use with Bio-Rad Chromo4 (Real-Time Detector), for use with Bio-Rad DNA Engine Opticon, for use with Bio-Rad Opticon 2, for use with Eppendorf® Mastercycler (ep realplex ), for use with Illumina Eco, for use with Roche LightCycler 1.5/2.0, for use with Roche LightCycler 480, for use with Roche LightCycler 96, for use with Roche LightCycler Nano, for use with Rotor-Gene®, for use with Smart Cycler®

detection method

SYBR® Green

detection method

SYBR® Green

detection method

SYBR® Green

detection method

SYBR® Green

manufacturer/tradename

Roche

manufacturer/tradename

Roche

manufacturer/tradename

Roche

manufacturer/tradename

Roche

concentration

2 ×

concentration

2 ×

concentration

2 ×

concentration

2 ×


Still not finding the right product?

Explore all of our products under KAPA SYBR® FAST


Kit Components Only

Product No.
Description

  • KAPA SYBR® FAST DNA Polymerase

  • Reaction buffer

  • dNTPs

  • SYBR® Green I dye

  • MgCl2 2.5 mM

  • ROX High Reference Dye (50X)

  • ROX Low Reference Dye (50X)

See All (8)

pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

STOT SE 2

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library