Skip to Content
Merck

Skip To

DTT-RO

Roche

DTT

crystalline powder, =97% (Ellman′s reagent), Mr 154.3

Synonym(s):

DL-Dithiothreitol, 1,4-dithiothreitol, cleland′s reagent, dithiothreitol, 1,4-, threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
HSCH2CH(OH)CH(OH)CH2SH
CAS Number:
Molecular Weight:
154.25
PubChem Substance ID:
UNSPSC Code:
12352200
Beilstein/REAXYS Number:
1719757
MDL number:
NACRES:
NA.54
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

1,4-Dithiothreitol

assay

97% (Ellman′s reagent)

form

crystalline powder

mol wt

Mr 154.3

reaction suitability

reagent type: reductant

packaging

pkg of 10 g (10708984001), pkg of 2 g (10197777001), pkg of 25 g (11583786001)

manufacturer/tradename

Roche

pH

5.1 (20 °C)

mp

41-44 °C (lit.)

storage temp.

2-8°C

SMILES string

O[C@H](CS)[C@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m1/s1

InChI key

VHJLVAABSRFDPM-QWWZWVQMSA-N

General description

1,4-dithiothreitol is commonly known as Cleland′s reagent. It confers protection to thiol groups and reduces disulfide bonds in peptides and proteins. It reduces disulfide bonds to sulfhydryl group and is commonly used in receptor studies, to determine the functional importance of disulfide bonds during receptor occupancy and functionality.[1] It also maintains monothiols completely in the reduced state.

Application

1,4-dithiothreitol is used in:
  • Isolation, purification and characterization of proteins and enzymes[2]
  • Measurement of enzyme activities (reactivation of enzymes)
  • Determination of disulfide groups in proteins and enzymes
  • DNA extraction prior to amplification[3]
An excellent reagent for maintaining SH groups in reduced state; quantitatively reduces disulfides. DTT is effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE. DTT can also be used for reducing the disulfide bridge of the cross-linker N,N′-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel. DTT is less pungent and is less toxic than 2-mercaptoethanol. Typically, a seven fold lower concentration of DTT (100 mM) is needed than is used for 2-mercaptoethanol (5% v/v, 700 mM).

Features and Benefits

Oxidized form: <2.5% (absorbance at 283nm)

Preparation Note

Storage conditions (working solution): A solution of DTT in Hepes buffer, pH 7.75 is stable for one week at 2 to 8 °C if the container is tightly sealed and the solution is protected from atmospheric oxygen by argon or nitrogen.

Other Notes

Enantiomers
Our DTT-preparation is optically inactive, i.e. it is the D,L-DTT.
For life science research only. Not for use in diagnostic procedures.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
3860-OPD0632D9779
DTT crystalline powder, =97% (Ellman&#8242;s reagent), Mr 154.3

Roche

DTT-RO

DTT

assay

97% (Ellman′s reagent)

assay

≥99.4%

assay

≥98% (HPLC), ≥99.0% (titration)

assay

≥98% (HPLC), ≥99% (titration)

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

2-8°C

form

crystalline powder

form

crystalline powder

form

powder

form

powder

pH

5.1 (20 °C)

pH

-

pH

-

pH

-

mp

41-44 °C (lit.)

mp

-

mp

41-44 °C (lit.)

mp

41-44 °C (lit.)

mol wt

Mr 154.3

mol wt

-

mol wt

-

mol wt

-


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



J Donaldson et al.
British journal of pharmacology, 87(1), 191-199 (1986-01-01)
1,4-Dithiothreitol (DTT; 1 mM, 30 min preincubation) produced a small, non-specific potentiation of spasmogenic activity in longitudinal muscle strips of guinea-pig small intestine. A direct comparison of contractile responses elicited by histamine and a range of H1- and non-H1-receptor agonists
Frans J L Robberts et al.
Journal of clinical microbiology, 42(4), 1505-1510 (2004-04-09)
Sequence analysis of Pneumocystis jiroveci internal transcribed spacer (ITS) regions has become an important epidemiological tool. The objectives of the present study were to investigate sequence variations in the ITS1-5.8S ribosomal DNA (rDNA)-ITS2 regions; determine the P. jiroveci genotypes present
Emanuelle Pascolo et al.
The Journal of biological chemistry, 277(18), 15566-15572 (2002-02-21)
Telomerase, a ribonucleoprotein acting as a reverse transcriptase, has been identified as a target for cancer drug discovery. The synthetic, non-nucleosidic compound, BIBR1532, is a potent and selective telomerase inhibitor capable of inducing senescence in human cancer cells (). In



Global Trade Item Number

SKUGTIN
1158378600104061838703668
1070898400104061838251800
1019777700104061838251770