Skip to Content
Merck

Skip To

TCTP14250

Polycarbonate Membrane Filter, 10.0 μm Pore Size

Isopore, filter diam. 142 mm, hydrophilic

Synonym(s):

Isopore Membrane Filter

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
10.0 μm pore size, 5-20 % porosity
Material:
plain filter, polycarbonate membrane, white filter
Feature:
hydrophilic
Diam.:
142 mm
Sterility:
non-sterile
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

plain filter, polycarbonate membrane, white filter

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Isopore, Millipore

parameter

140 °C max. temp., 69 L/min-cm2-psi air flow rate, ~900 mL/min-cm2 water flow rate at 10 psi (typical results)

diam.

142 mm

filter diam.

142 mm

thickness

16 μm

gravimetric extractables

<1%

refractive index

n/D 1.6

matrix

Isopore

pore size

10.0 μm pore size, 5-20 % porosity

shipped in

ambient

General description

Filter Code: TCTP
Filter Type: Screen filter
The Isopore membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.

Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety

Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays

Application

  • Chemotaxis
  • Bioassays
  • Cytology
  • Air monitoring
  • Microplastics analysis

Legal Information

ISOPORE is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Possible reduction in flow rates may occur after autoclaving.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
TCTP01300TCTP02500TCTP04700
parameter

140 °C max. temp., ~900 mL/min-cm2 water flow rate at 10 psi (typical results), 69 L/min-cm2-psi air flow rate

parameter

140 °C max. temp., 69 L/min-cm2-psi air flow rate, ~900 mL/min-cm2 water flow rate at 10 psi (typical results)

parameter

140 °C max. temp., 69 L/min-cm2-psi air flow rate, ~900 mL/min-cm2 water flow rate at 10 psi (typical results)

parameter

140 °C max. temp., 69 L/min-cm2-psi air flow rate, ~900 mL/min-cm2 water flow rate at 10 psi (typical results)

pore size

10.0 μm pore size, 5-20 % porosity

pore size

10.0 μm pore size, 5-20 % porosity

pore size

10.0 μm pore size, 5-20 % porosity

pore size

10.0 μm pore size, 5-20 % porosity

diam.

142 mm

diam.

13 mm

diam.

25 mm

diam.

47 mm

material

plain filter, white filter, polycarbonate membrane

material

plain filter, polycarbonate membrane, white filter

material

plain filter, polycarbonate membrane, white filter

material

plain filter, polycarbonate membrane, white filter

manufacturer/tradename

Isopore, Millipore

manufacturer/tradename

Isopore, Millipore

manufacturer/tradename

Isopore, Millipore

manufacturer/tradename

Isopore, Millipore

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile


Still not finding the right product?




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
TCTP1425004053252724978

Questions

Reviews

No rating value

Active Filters