Skip to Content
Merck

Skip To

MPMES

MES SDS running buffer powder for mPAGE® Bis-Tris gels

pack of 5 ea (Each packet makes 1 liter of running buffer)

Synonym(s):

Bis-Tris running buffer, SDS-PAGE running buffer, running buffer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.56
UNSPSC Code:
12161703
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

packaging

pack of 5 ea (Each packet makes 1 liter of running buffer)

General description

MES SDS running buffer powder for mPAGEBis-Tris gels. 5 packets. Each paclet makes 1 liter of running buffer.
  • Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Application

MES SDS buffer is formulated for running mPAGE Bis-Tris gels. This buffer is recommended for separating small- and medium- sized proteins.

Reconstitute 1 pouch of buffer powder with 1000 ml deionized water to make 1X running buffer. Do not use acid or base to adjust the pH.

  • Western Blotting

Legal Information

MPAGE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MPM0PSMPTRBPPB019
form

powder

form

powder

form

powder

form

powder

packaging

pack of 5 ea (Each packet makes 1 liter of running buffer)

packaging

pack of 5 ea (Each packet makes 1 liter of running buffer)

packaging

pack of 10 ea (Each packets makes 1 liter of transfer buffer at 1X)

packaging

-


Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - STOT RE 1

target_organs

Thyroid



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MPMES04054839761744

Questions

Reviews

No rating value

Active Filters