Skip to Content
Merck

Skip To

MABE282

Anti-c-Myc Tag Antibody

mouse monoclonal, 9E10

Synonym(s):

Myc proto-oncogene protein, Class E basic helix-loop-helix protein 39, HLHe39, Proto-oncogene c-Myc, Transcription factor p64

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Clone:
9E10, monoclonal
Species reactivity:
human
Application:
ChIP, WB
Citations:
23
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-c-Myc Antibody, clone 9E10, clone 9E10, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9E10, monoclonal

species reactivity

human

technique(s)

ChIP: suitable, western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

General description

c-Myc (bHLHe39) is a transcription factor that regulates the expression of multiple genes involved in apoptosis, cell differentiation and cell proliferation. Active c-Myc is coupled to the MAX protein via c-Myc’s C-terminal helix-loop-helix leucine zipper (bHLHLZ) domain. The c-Myc-MAX heterodimer also interacts with a number of other transcription-related proteins including TRRAP, p107 and Miz-1, and c-Myc-MAX may indirectly regulate histone acetylases involved in chromatin remodeling to increase accessibility of transcriptional complexes to target DNA sequences. The expression of c-Myc is controlled by growth factors. Several studies have reported abnormal expression of c-Myc in various cancers including Burkitt’s lymphoma.
~51 kDa observed. Uniprot describes two isoforms at ~49 kDa and ~51 kDa

Immunogen

Linear peptide corresponding to human c-Myc.

Application

Anti-c-Myc Antibody, clone 9E10 is a Mouse Monoclonal Antibody for detection of c-Myc also known as Myc proto-oncogene protein, HLHe39, Transcription factor p64 & has been validated in WB, ChIP.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Western Blot Analysis: A representative lot from an independent laboratory detected c-Myc in various cell lysates (Evan, G. I., et al. (1985). Mol Cell Biol. 5(12):3610-3616).

Chromatin Immunoprecipitation Analysis: A representative lot from an independent laboratory detected c-Myc in ChIP (Sanyal, K., et al. (2004). Proc Natl Acad Sci USA. 101(31):11374-11379).

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HeLa cell lysate
Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected c-Myc in 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB4700448WH0004609M2SAB4501941
antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

affinity isolated antibody

Gene Information

human ... MYC(4609)

Gene Information

human ... MYC(4609)

Gene Information

human ... MYC(4609)

Gene Information

human ... MYC(4609)

biological source

mouse

biological source

mouse

biological source

mouse

biological source

rabbit

clone

9E10, monoclonal

clone

9E10, monoclonal

clone

1G7, monoclonal

clone

polyclonal

species reactivity

human

species reactivity

human, fusion proteins in all species

species reactivity

human

species reactivity

rat, mouse, human

technique(s)

ChIP: suitable, western blot: suitable

technique(s)

flow cytometry: suitable

technique(s)

immunofluorescence: suitable, immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, indirect ELISA: suitable, western blot: 1-5 μg/mL

technique(s)

ELISA: 1:40000, immunohistochemistry: 1:50-1:100, western blot: 1:500-1:1000


Still not finding the right product?

Explore all of our products under Anti-c-Myc Antibody, clone 9E10

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

New Products: Antibodies, Assays, Small Molecules, Inhibitors, and Proteins


Samuel Rommelaere et al.
Cell reports, 27(3), 886-899 (2019-04-18)
In ectotherms, increased ambient temperature requires the organism to consume substantial amounts of energy to sustain a higher metabolic rate, prevent cellular damage, and respond to heat stress. Here, we identify a heat-inducible apolipoprotein required for thermal acclimation in Drosophila.
Iga Dalidowska et al.
International journal of molecular sciences, 23(13) (2022-07-10)
Mononegavirales is an order of viruses with a genome in the form of a non-segmented negative-strand RNA that encodes several proteins. The functional polymerase complex of these viruses is composed of two proteins: a large protein (L) and a phosphoprotein
Iga Dalidowska et al.
International journal of molecular sciences, 22(4) (2021-03-07)
Adenovirus infections tend to be mild, but they may pose a serious threat for young and immunocompromised individuals. The treatment is complicated because there are no approved safe and specific drugs for adenovirus infections. Here, we present evidence that 17-(Allylamino)-17-demethoxygeldanamycin



Global Trade Item Number

SKUGTIN
MABE28204053252549298

Questions

Reviews

No rating value

Active Filters