Skip to Content
Merck

Skip To

HATF04700

Immobilon® -NC Nitrocellulose Membrane

greener alternative

100 discs, 47 mm, 0.45 µm pore size, Triton-free, nitrocellulose-based transfer membrane

Synonym(s):

Western blotting membrane, blotting membrane, nitrocellulose transfer membrane, transfer membrane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
Material:
mixed cellulose esters (MCE) membrane (surfactant-free), nitrocellulose membrane, plain filter, white filter
Detection method:
chemiluminescent, colorimetric, fluorometric, radioactive
Pore size:
0.45 μm pore size, 79 % porosity
Compatibility:
for use with Amido black, for use with CPTS, for use with Colloidal gold, for use with India ink, for use with Ponceau-S red
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Immobilon®-NC Transfer Membrane, 47 mm diameter disc, 0.45 µm pore size, Triton-free, mixed cellulose esters transfer membrane

Quality Level

material

mixed cellulose esters (MCE) membrane (surfactant-free), nitrocellulose membrane, plain filter, white filter

feature

hydrophilic

manufacturer/tradename

Immobilon®

sustainability

Greener Alternative Product

parameter

4 L/min-cm2 air flow rate, 60 mL/min-cm2 water flow rate, 75 °C max. temp.

technique(s)

western blot: suitable

filter diam.

47 mm

thickness

150 μm

refractive index

n/D 1.51

matrix

Immobilon®

pore size

0.45 μm pore size, 79 % porosity

capacity

117 μg/cm2 adsorption capacity (insulin), 160 μg/cm2 adsorption capacity (BSA), 259 μg/cm2 adsorption capacity (goat IgG)

bubble point

≥2.1 bar, air with water at 23 °C

compatibility

for use with Amido black, for use with CPTS, for use with Colloidal gold, for use with India ink, for use with Ponceau-S red

detection method

chemiluminescent, colorimetric, fluorometric, radioactive

greener alternative category

shipped in

ambient

General description

Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.

Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Filter Code: HATF
Primary Binding Mechanisms: Electrostatic, hydrophobic
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner & outer boxes containing the membrane discs are made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

  • Biological solutions
  • Cell contact
  • Very small volumes requiring surfactant-free surfaces
  • Isolation of virus-like particles in wastewater
  • Microplastics analysis grade water
  • Nucleic acid binding, including eDNA

Legal Information

Immobilon is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HATF00010HATF14250HATF01300
pore size

0.45 μm pore size, 79 % porosity

pore size

0.45 μm pore size

pore size

0.45 μm pore size, 79 % porosity

pore size

0.45 μm pore size, 79 % porosity

Quality Level

400

Quality Level

400

Quality Level

400

Quality Level

400

material

mixed cellulose esters (MCE) membrane (surfactant-free), plain filter, nitrocellulose membrane, white filter

material

mixed cellulose esters (MCE) membrane, nitrocellulose membrane, plain filter, white filter

material

mixed cellulose esters (MCE) membrane (surfactant-free), plain filter, white filter

material

mixed cellulose esters (MCE) membrane (surfactant-free), plain filter, white filter

compatibility

for use with Amido black, for use with Colloidal gold, for use with Ponceau-S red, for use with CPTS, for use with India ink

compatibility

for use with Amido black, for use with CPTS, for use with Colloidal gold, for use with India ink, for use with Ponceau-S red

compatibility

for use with Amido black, for use with CPTS, for use with Colloidal gold, for use with India ink, for use with Ponceau-S red

compatibility

for use with Amido black, for use with CPTS, for use with Colloidal gold, for use with India ink, for use with Ponceau-S red

detection method

chemiluminescent, fluorometric, radioactive, colorimetric

detection method

chemiluminescent, colorimetric, fluorometric, radioactive

detection method

colorimetric, fluorometric, radioactive

detection method

colorimetric, fluorometric, radioactive

manufacturer/tradename

Immobilon®

manufacturer/tradename

Immobilon®

manufacturer/tradename

Immobilon®, Millipore

manufacturer/tradename

Immobilon®, Millipore


Still not finding the right product?

Explore all of our products under Immobilon®-NC Transfer Membrane


pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Sol. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
HATF0470008436037123795

Questions

Reviews

No rating value

Active Filters