Skip to Content
Merck

Skip To

GTTP02500

Polycarbonate Membrane Filter, 0.22 μm Pore Size

greener alternative

Isopore, filter diam. 25 mm, hydrophilic, white

Synonym(s):

Hydrophilic polycarbonate membrane filter, Isopore Membrane Filter, Polycarbonate membrane filter discs

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
eCl@ss:
32031602
Pore size:
0.22 μm pore size, 13.8 % porosity
Material:
plain filter, polycarbonate membrane, white filter
Feature:
hydrophilic
Diam.:
25 mm
Sterility:
non-sterile
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

plain filter, polycarbonate membrane, white filter

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Isopore, Millipore

sustainability

Greener Alternative Product

parameter

0.007 L/min-cm2-psi air flow rate, 140 °C max. temp., 3.36 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi), ~16 mL/min-cm2 water flow rate at 10 psi (typical results)

diam.

25 mm

filter diam.

25 mm

thickness

25 μm

gravimetric extractables

<1%

color

white

refractive index

n/D 1.6

matrix

Isopore

pore size

0.22 μm pore size, 13.8 % porosity

bubble point

≥5.2 bar, air with water at 23 °C

greener alternative category

shipped in

ambient

General description

Filter Code: GTTP
Filter Type: Screen filter
The Isopore membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.

Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety

Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
We are committed to bringing you Greener Alternative Products, which have been re-engineered to enable more sustainable research solutions. The inner & outer boxes containing the membrane discs are made from bio-circular material allocated using the ISCC mass balance approach. Click here for more information.

Application

  • Chemotaxis
  • Bioassays
  • Cytology
  • Air monitoring
  • SEM analysis
  • Sterility testing
  • Microplastics analysis
  • Microplastics analysis grade water

Legal Information

ISOPORE is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Possible reduction in flow rates may occur after autoclaving.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ATTP02500GTTP04700GTTP01300
manufacturer/tradename

Isopore, Millipore

manufacturer/tradename

Isopore, Millipore

manufacturer/tradename

Isopore, Millipore

manufacturer/tradename

Isopore, Millipore

diam.

25 mm

diam.

25 mm

diam.

47 mm

diam.

13 mm

pore size

0.22 μm pore size, 13.8 % porosity

pore size

0.8 μm pore size, 5-20 % porosity

pore size

0.22 μm pore size, 13.8 % porosity

pore size

0.22 μm pore size, 13.8 % porosity

material

plain filter, white filter, polycarbonate membrane

material

plain filter, polycarbonate membrane, white filter

material

plain filter, polycarbonate membrane, white filter

material

plain filter, polycarbonate membrane, white filter

parameter

0.007 L/min-cm2-psi air flow rate, 3.36 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi), 140 °C max. temp., ~16 mL/min-cm2 water flow rate at 10 psi (typical results)

parameter

125 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi), 140 °C max. temp., 4.08 L/min-cm2-psi air flow rate, ~85 mL/min-cm2 water flow rate at 10 psi (typical results)

parameter

0.007 L/min-cm2-psi air flow rate, 140 °C max. temp., 3.36 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi), ~16 mL/min-cm2 water flow rate at 10 psi (typical results)

parameter

0.007 L/min-cm2-psi air flow rate, 140 °C max. temp., 3.36 mL/min-cm2-psi water flow rate (maximum flow rate per 1.0 psi), ~16 mL/min-cm2 water flow rate at 10 psi (typical results)

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile


Still not finding the right product?




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. I use it often to filter cells. I was just wondering if it matters which side of the filter I use? Are we supposed to use it the way they come in the box? So that the top of the filter is always used as the part to put the cell suspension on?

    1 answer
    1. Isopore membranes should be handled carefully using appropriate forceps, such as Millipore forceps (Catalog No. XX6200006P). Avoid touching the membrane with fingers. Membranes should be used with the shiny side up.

      Helpful?

Reviews

No rating value

Active Filters