Skip to Content
Merck

Skip To

ABC11

Anti-Bax (NT) Antibody

from rabbit

Synonym(s):

BCL2-associated X protein, Bcl-2-like protein 4, apoptosis regulator BAX

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, IHC, IP, WB
Citations:
17
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), feline (based on 100% sequence homology), canine (based on 100% sequence homology), chimpanzee (based on 100% sequence homology), rhesus monkey (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable, immunohistochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BAX(581)

General description

BAX (BCL2-associated X protein) belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. BAX protein forms a heterodimer with BCL2, and functions as an apoptotic activator. This protein is reported to interact with, and increase the opening of, the mitochondrial voltage-dependent anion channel (VDAC), which leads to the loss in membrane potential and the release of cytochrome c. The expression of this gene is regulated by the tumor suppressor P53 and has been shown to be involved in P53-mediated apoptosis. Multiple alternatively spliced transcript variants, which encode different isoforms, have been reported for this gene.
The previously assigned protein identifier Q07815 has been merged into Q07812. Full details can be found on the UniProt database.
~ 21 kDa

Immunogen

KLH-conjugated linear peptide corresponding to human BAX at and around the N-terminus.

Application

Anti-Bax Antibody is an antibody against Bax (N-terminus) for use in WB, IP, IC, IH.
Immunoprecipitation Analysis: 10 µg from a previous lot immunoprecipitated BAX from 500 µg of HL60 cell lysate.

Immunohistochemistry Analysis: 1:300 dilution from a previous lot detected BAX in colorectal and ductal carcinoma tissue.

Immunocytochemistry Analysis: 1:500 dilution from a previous lot detected BAX in MCF7 cells.

Biochem/physiol Actions

Rat (90%), Mouse (86%).
This antibody recognizes BAX.

Physical form

Format: Purified

Analysis Note

Evaluated by Western Blot in HL60 cell lysate.

Western Blot Analysis: 0.125 µg/ml of this antibody detected BAX on 10 µg of HL60 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 06-499

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB5700071SAB5700840SAB5700056
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, immunohistochemistry: suitable, western blot: suitable

technique(s)

immunohistochemistry: 1:100-1:200, immunoprecipitation (IP): 1:50-1:200, western blot: 1:500-1:2000

technique(s)

western blot: 1:500-1:2000

technique(s)

immunofluorescence: 1:50-1:100, western blot: 1:500-1:1000


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Adeline Y Robin et al.
Structure (London, England : 1993), 26(10), 1346-1359 (2018-08-21)
BAX and BAK are essential mediators of intrinsic apoptosis that permeabilize the mitochondrial outer membrane. BAX activation requires its translocation from cytosol to mitochondria where conformational changes cause its oligomerization. To better understand the critical step of translocation, we examined
Teruyoshi Saito et al.
Oncology reports, 29(1), 335-342 (2012-11-06)
Hyaluronan (HA), a major component of the extracellular matrix (ECM), is synthesized by HA synthase (HAS) 1, HAS2 and HAS3 and is intricately involved
Dan-Dan Sun et al.
Journal of dental sciences, 18(1), 310-321 (2023-01-17)
Periodontitis is a prevalent infectious inflammatory disease. Growing evidence has revealed important roles for circular RNAs (circRNAs) and circRNA sponge activity in periodontitis. Here, we elucidated the precise part of circ_0097010 in periodontitis pathogenesis. Human periodontal ligament cells (hPDLCs) were



Global Trade Item Number

SKUGTIN
ABC1104053252331749

Questions

Reviews

No rating value

Active Filters