Skip to Content
Merck

Skip To

8.07679

Styrene

(stabilised) for synthesis

Synonym(s):

Styrene, Phenylethylene, Vinylbenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H8
CAS Number:
Molecular Weight:
104.15
UNSPSC Code:
12352005
EC Index Number:
202-851-5
NACRES:
NA.22
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

6 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

480 °C

potency

2650 mg/kg LD50, oral (Rat)

expl. lim.

1.1-8.9 % (v/v)

bp

145 °C/1013 hPa

mp

-31 °C

transition temp

flash point 31 °C

solubility

0.24 g/L

density

0.906 g/cm3 at 20 °C

SMILES string

c1(ccccc1)C=C

InChI

1S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7H,1H2

InChI key

PPBRXRYQALVLMV-UHFFFAOYSA-N

General description

(stabilised) for synthesis

Application

Styrene (stabilised) for synthesis. CAS 100-42-5, EC Number 202-851-5, chemical formula C₆H₅CHCH₂.

Features and Benefits

Saturation Concentration in Air: 25.6 g/m3 (20 °C)

Analysis Note

Assay (GC, area%)≥ 99.0 % (a/a)Density (d 20 °C/ 4 °C)0.905 - 0.907Identity (IR)passes testThe material is stabilised with 4-tert-Butylpyrocatechol. At time of analytical testing a content of >10 ppm stabiliser was determined.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S4972PHR3500907146
mp

-31 °C

mp

−31 °C (lit.)

mp

-

mp

−31 °C (lit.)

density

0.906 g/cm3 at 20 °C

density

0.906 g/mL at 25 °C

density

-

density

0.906 g/mL at 25 °C

bp

145 °C/1013 hPa

bp

145-146 °C (lit.)

bp

-

bp

145-146 °C (lit.)

Quality Level

200

Quality Level

200

Quality Level

300

Quality Level

-

solubility

0.24 g/L

solubility

-

solubility

-

solubility

-

autoignition temp.

480 °C

autoignition temp.

914 °F

autoignition temp.

-

autoignition temp.

914 °F


Still not finding the right product?

Explore all of our products under Styrene


signalword

Danger

target_organs

hearing organs, Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

89.6 °F - closed cup

flash_point_c

32.0 °C - closed cup

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
807679100004022536396972
807679000004022536396958
807679800404065267525501
807679800504054839985393
807679250004022536396989
807679010004022536396965
807679999904022536397009

Questions

1–4 of 4 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. The styrene is stabilized. What substance is the styrene stabilized with?

    1 answer
    1. As mentioned in the 'DESCRIPTION' section under "Analysis Note', Styrene is stabilized with 4-tert-Butylpyrocatechol.

      Helpful?

  4. is 8.07679 Styrene CAS Number: 100-42-5 a MONOMER?

    1 answer
    1. This product is a monomeric styrene with a molecular weight of 104.15 g/mol.

      Helpful?

Reviews

No rating value

Active Filters