Skip to Content
Merck

Skip To

1.99036

Buffer solution

(citric acid/sodium hydroxide), traceable to SRM from NIST and PTB pH 6.00 (25°C) Certipur®

Synonym(s):

Buffer solution pH 6.0 (25 °C in H2O) (citric acid/sodium hydroxide)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

NACRES:
NA.25
UNSPSC Code:
12161703
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Level

product line

Certipur®

pH

6.0 (25 °C in H2O)

density

1.01 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

The citric acid/sodium hydroxide buffer solution is gravimetrically prepared at a registered calibration laboratory with high-quality reagents and water. Its pH value of 6.00 at 25°C is measured using a combined glass electrode following a 5-point calibration method. The measurement is made following DIN 19268 using reference buffer solutions compliant to DIN 19266, NIST, IUPAC, Ph. Eur, and USP. The buffer pH depends on the temperature and therefore a constant temperature must be maintained during its use.

Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.

Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

The buffer is used as a standard to calibrate pH measuring instruments or pH electrodes and as a control to measure the pH of other solutions.

Features and Benefits

  • Measured and qualified at an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Traceable to secondary reference standard from NIST and primary certified reference standard from PTB
  • Available in a secure 500 mL plastic bottle and in 4 L innovative and sustainable Titripac® PE/Aluminum packaging for a reduced waste generation
  • Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective audit
  • Product label follows the ISO Guide 31 and contains information about ordering number, article description, lot number, safety data, and expiry date
  • Packaging material subjected to comprehensive stability studies as per ISO Guide 35

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Titripac is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.990641.094451.09446
format

mixture

format

mixture

format

mixture

format

mixture

product line

Certipur®

product line

Certipur®

product line

Certipur®

product line

Certipur®

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

pH

6.0 (25 °C in H2O)

pH

4.0 (25 °C in H2O)

pH

4.0 (25 °C in H2O)

pH

5.0 (25 °C in H2O)

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C


Still not finding the right product?

Explore all of our products under Buffer solution


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
199036050004027269386832
199036400004027269386849

Questions

Reviews

No rating value

Active Filters