If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf
Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product Name
ortho-Phosphoric acid 85%, Suprapur®
vapor density
3.4 (vs air)
Quality Level
vapor pressure
2.2 mmHg ( 20 °C), 5 mmHg ( 25 °C)
description
for inorganic trace analysis
assay
85%
form
liquid
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
pH
<0.5 (20 °C, 100 g/L in H2O)
kinematic viscosity
30.5 cSt(20 °C)
bp
158 °C (lit.)
mp
~40 °C (lit.)
density
1.685 g/mL at 25 °C (lit.)
greener alternative category
storage temp.
no temp limit
SMILES string
OP(O)(O)=O
InChI
1S/H3O4P/c1-5(2,3)4/h(H3,1,2,3,4)
InChI key
NBIIXXVUZAFLBC-UHFFFAOYSA-N
General description
Application
- Development of green micellar HPLC-DAD method for simultaneous determination of some sulbactam combinations used in COVID-19 regimen: This study presents a green analytical methodology utilizing micellar high-performance liquid chromatography with diode-array detection (HPLC-DAD). It is specifically employed for the simultaneous determination of sulbactam and other compounds in COVID-19 treatment regimens, showcasing an application of ortho-Phosphoric acid in adjusting the pH of the mobile phase to enhance analytical performance and separation efficiency (Ragab GH, Saleh HM, Abdulla NM, Bahgat EA, 2023).
Analysis Note
Chloride (Cl): ≤ 500 ppb
Fluoride (F): ≤ 500 ppb
Nitrate (NO₃): ≤ 500 ppb
Sulphate (SO₄): ≤ 10 ppm
Ag (Silver): ≤ 20 ppb
Al (Aluminium): ≤ 20 ppb
As (Arsenic): ≤ 25 ppb
Au (Gold): ≤ 20 ppb
Ba (Barium): ≤ 20 ppb
Be (Beryllium): ≤ 50 ppb
Bi (Bismuth): ≤ 50 ppb
Ca (Calcium): ≤ 50 ppb
Cd (Cadmium): ≤ 20 ppb
Co (Cobalt): ≤ 20 ppb
Cr (Chromium): ≤ 50 ppb
Cu (Copper): ≤ 25 ppb
Fe (Iron): ≤ 200 ppb
Hg (Mercury): ≤ 2 ppb
K (Potassium): ≤ 50 ppb
Li (Lithium): ≤ 20 ppb
Mg (Magnesium): ≤ 20 ppb
Mn (Manganese): ≤ 20 ppb
Mo (Molybdenum): ≤ 20 ppb
Na (Sodium): ≤ 100 ppb
Ni (Nickel): ≤ 25 ppb
Pb (Lead): ≤ 20 ppb
Pt (Platinum): ≤ 100 ppb
Sb (Antimony): ≤ 800 ppb
Sn (Tin): ≤ 50 ppb
Sr (Strontium): ≤ 20 ppb
Tl (Thallium): ≤ 20 ppb
V (Vanadium): ≤ 20 ppb
Zn (Zinc): ≤ 25 ppb
Zr (Zirconium): ≤ 50 ppb
The actual values are subject to unavoidable systematic
variations in this concentration range.
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| assay 85% | assay - | assay ≥99% | assay ≥85% (T) |
| Quality Level 100 | Quality Level 500 | Quality Level - | Quality Level 200 |
| form liquid | form liquid | form - | form viscous liquid |
| storage temp. no temp limit | storage temp. 2-30°C | storage temp. - | storage temp. - |
| pH <0.5 (20 °C, 100 g/L in H2O) | pH - | pH - | pH - |
| density 1.685 g/mL at 25 °C (lit.) | density 1.053 g/cm3 at 20 °C | density 1.685 g/mL at 25 °C (lit.) | density 1.685 g/mL at 25 °C (lit.) |
Still not finding the right product?
Explore all of our products under ortho-Phosphoric acid 85%
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B
wgk
WGK 1
flash_point_f
does not flash
flash_point_c
does not flash
Storage Class
8B - Non-combustible corrosive hazardous materials
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 1005520500 | 04027269294618 |
| 1005522500 | 04027269301170 |
| 1005521000 | 04022536007724 |
| 1005520250 | 04022536007717 |
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-



