Skip to Content
Merck

Skip To

F-047

Flubromazolam solution

1.0 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C17H12BrFN4
CAS Number:
Molecular Weight:
371.21
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Pszichotróp anyag / Psychotropic Substance (Hungary), 78/2022. (XII. 28.) BM rendelet, kontrollierte Droge in Deutschland

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Fc1c(cccc1)C2=NCc3[n](c(nn3)C)c4c2cc(cc4)Br

InChI

1S/C17H12BrFN4/c1-10-21-22-16-9-20-17(12-4-2-3-5-14(12)19)13-8-11(18)6-7-15(13)23(10)16/h2-8H,9H2,1H3

InChI key

VXGSZBZQCBNUIP-UHFFFAOYSA-N

General description

This Certified Spiking Solution® is suitable for testing flubromazolam by HPLC, GC/MS or LC-MS/MS methods in forensic analysis, clinical toxicology, urine drug testing, and cannabis potency or impurity profiling testing applications.

Flubromazolam, a psychoactive compound, is a triazole analog of the designer benzodiazepine, flubromazepam.

Application

The Certified Reference Material (CRM) in solution can also be used as follows:

  • Residue analysis of nine designer benzodiazepines (DBZDs) in serum samples after their liquid-liquid extraction by non-aqueous capillary electrophoresis coupled with tandem mass spectrometry(MS/MS)
  • Simultaneous determination of 11 designer benzodiazepines from urine samples by liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • Ultra high-performance liquid chromatography-time-of-flight mass spectrometry (UHPLC-TOF-MS) method based separation and quantification of designer benzodiazepines from serum samples following their liquid-liquid extraction
  • Combination of parallel artificial liquid membrane extraction (PALME) with UHPLC-MS/MS to determine benzodiazepines, designer benzodiazepines, and z-hypnotics from whole blood samples
  • Urine analysis for the testing of 28 designer benzodiazepines reversed phase-LC combined with high-resolution mass spectrometry (HRMS)

Features and Benefits

  • Fully characterized under ISO/IEC 17025 and ISO 17034 accreditation
  • Accompanied with a comprehensive Certificate of Analysis (CoA) with data on stability, homogeneity, accuracy of concentration, uncertainty, and traceability
  • Rigorously tested through real-time stability studies to ensure accuracy and shelf life
  • Gravimetrically prepared using qualified precision balances to ensure minimal uncertainty
  • Flame sealed under argon into ampoules for long-term shelf life
  • Offered in a convenient, DEA-exempt format to improve laboratory efficiency

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Urine analysis of 28 designer benzodiazepines by liquid chromatography-high-resolution mass spectrometry
Bergstrand MP, et al.
Clinical mass spectrometry, 10, 25-32 (2018)



Global Trade Item Number

SKUGTIN
F-047-1ML04061838054913

Questions

Reviews

No rating value

Active Filters