Skip to Content
Merck

Skip To

D-078

11-Deoxycortisol-D5 (2,2,4,6,6-D5)

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

11-Deoxycortisol-D5 (2,2,4,6,6-D5) solution, 11-Desoxycortisol-D5 (2,2,4,6,6-D5), Cortodoxone-D5 (2,2,4,6,6-D5)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21D5H25O4
CAS Number:
Molecular Weight:
351.49
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-659-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

[2H]C1=C2C([2H])([2H])C[C@@H]3[C@H](CC[C@@]4(C)[C@H]3CC[C@]4(O)C(=O)CO)[C@@]2(C)CC([2H])([2H])C1=O

InChI

1S/C21H30O4/c1-19-8-5-14(23)11-13(19)3-4-15-16(19)6-9-20(2)17(15)7-10-21(20,25)18(24)12-22/h11,15-17,22,25H,3-10,12H2,1-2H3/t15-,16+,17+,19+,20+,21+/m1/s1/i3D2,5D2,11D

InChI key

WHBHBVVOGNECLV-NGKMQDFHSA-N

General description

An internal standard for 11-deoxycortisol, also known as cortodoxone. An immediate precursor to cortisol, 11-deoxycortisol is measured in whole blood by LC-MS/MS for newborn screening of congenital adrenal hyperplasia (CAH) and in serum or plasma by LC-MS/MS for determination of several diseases including Cushing′s syndrome, pituitary tumor, and Addison′s disease.

Legal Information

Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
D-078-1ML04061833556856

Questions

Reviews

No rating value

Active Filters