Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
grade
certified reference material
Quality Level
form
liquid
feature
SNAP-N-SPIKE®, SNAP-N-SHOOT®
packaging
ampule of 1 mL
manufacturer/tradename
Cerilliant®
drug control
Narcotic Licence Schedule A (Switzerland); estupefaciente (Spain); Decreto Lei 15/93: Tabela IB (Portugal)
concentration
100 μg/mL in acetonitrile
technique(s)
gas chromatography (GC): suitable, liquid chromatography (LC): suitable
application(s)
forensics and toxicology
format
single component solution
storage temp.
−20°C
SMILES string
[2H]C([2H])([2H])N1[C@H]2CC[C@@H]1C(C(C2)OC(=O)c3ccccc3)C(=O)OC([2H])([2H])C([2H])([2H])[2H]
InChI
1S/C18H23NO4/c1-3-22-18(21)16-14-10-9-13(19(14)2)11-15(16)23-17(20)12-7-5-4-6-8-12/h4-8,13-16H,3,9-11H2,1-2H3/t13-,14+,15?,16?/m0/s1/i1D3,2D3,3D2
InChI key
NMPOSNRHZIWLLL-WVIYWTTLSA-N
General description
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable | technique(s) gas chromatography (GC): suitable, liquid chromatography (LC): suitable |
| form liquid | form liquid | form liquid | form liquid |
| format single component solution | format single component solution | format single component solution | format single component solution |
| Quality Level 300 | Quality Level 300 | Quality Level 300 | Quality Level 300 |
| grade certified reference material | grade certified reference material | grade certified reference material | grade certified reference material |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
35.6 °F - closed cup
flash_point_c
2 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| C-024-1ML | 04061833444481 |





