Skip to Content
Merck

Skip To

D8906

Dextran sulfate sodium salt from Leuconostoc spp.

Molecular Biology, average Mw >500,000 (dextran starting material), contains 0.5-2% phosphate buffer

Synonym(s):

6-O-(6-O-beta-D-Glucopyranosyl-beta-D-glucopyranosyl)-D-glucose, Manninotriose

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.31
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Level

grade

Molecular Biology

form

powder

mol wt

average Mw >500,000 (dextran starting material)

contains

0.5-2% phosphate buffer

composition

Sulfur Content, 16.0-19.0%

technique(s)

FISH: suitable, FTIR: suitable

color

off-white

useful pH range

6.0-8 (10 g/L)

solubility

H2O: 100 mg/mL

suitability

suitable for marker for electrophoresis, suitable for nucleic acid hybridization

application(s)

genomic analysis
sample preparation

General description

Dextran sulfate sodium (DSS) is a synthetic sulfated derivative of dextran, a complex polysaccharide composed of glucose molecules. DSS is characterized as a biocompatible, highly branched polysaccharide with 1–6 and 1–4 glycosidic linkages, featuring approximately 2.3 sulfate groups per glucosyl unit. Dextran sulfate is a sulfated polysaccharide with a variable molecular weight ranging from 5 to 1,400 kDa. It finds extensive use in molecular biology and biochemical research, with a particular emphasis on gastrointestinal investigations.DSS is widely recognized for its ability to induce colonic inflammation in mouse models of inflammatory bowel disease (IBD). It is widely recognized for its capacity to induce colonic inflammation in mouse models of inflammatory bowel disease (IBD) by creating models of acute, chronic, and relapsing IBD, by adjusting its concentration and administration frequency. Furthermore, Dextran sulfate is applied in various molecular biology techniques, such as precipitating lipoproteins, accelerating DNA probe hybridization, and facilitating ribosomal isolations. Notably, DSS serves as a valuable cross-linker for chitosan, allowing for the creation of microparticles and nanoparticles used in controlled drug delivery systems. Beyond these applications, DSS plays a central role in diverse research areas, particularly immunology and virology, where it actively modulates immune responses, enhances humoral immunity, influences B-lymphocyte activation, and affects thymocyte reactivity to lectins.

Application

Dextran sulfate has many applications and when performing nucleic acid hybridizations, may be used as an accelerant to decrease the time required for probe annealing. Dextran sulfate may be included in the hybridization buffer when the rate of hybridization is a limiting factor in detecting rare sequences or it may be included in the hybridization buffer when a limited amount of probe is available. Dextran sulfate functions as an accelerant by excluding the nucleic acid from the volume of solution occupied by the polymer, thereby, increasing the effective probe concentration.
Tested for suitability in nucleic acid hybridizations
Dextran sulfate has been used

  • as a molecular weight marker to assess the purity and homogeneity of the purified polysaccharides by gel electrophoresis
  • as a component of the hybridization buffer in fluorescence in situ hybridization (FISH) protocol for DNA detection and analysis[1]

Biochem/physiol Actions

Dextran sulfate, being a sulfated polysaccharide, consistently triggers experimental acute and chronic colitis. It is believed to initiate mucosal injury and inflammation, primarily through a direct toxic impact on epithelial cells. This initial effect activates macrophages and T lymphocytes, ultimately leading to cytokine-mediated cytotoxicity.

Features and Benefits

  • Suitable for Nucleic acid hybridization & Molecular Biology
  • Dextran Sulfate with an average molecular weight of >500,000
  • Sulfur Content of 16.0-19.0 %
  • Versatile and adaptable for wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.
To gain a comprehensive understanding of our extensive range of Dextrans for your research, we encourage you to visit our Carbohydrates Category page.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
52423D6924265152-M
technique(s)

FISH: suitable, FTIR: suitable

technique(s)

-

technique(s)

-

technique(s)

-

biological source

synthetic

biological source

(Leuconostoc spp)

biological source

bacterial (Leuconostoc mesenteroides)

biological source

-

grade

Molecular Biology

grade

-

grade

-

grade

Molecular Biology

Quality Level

300

Quality Level

100

Quality Level

200

Quality Level

100

solubility

H2O: 100 mg/mL

solubility

H2O: 100 mg/mL, clear, colorless to faintly greenish-yellow

solubility

H2O: 100 mg/mL

solubility

water: 5 mg/mL

form

powder

form

powder

form

powder

form

solid


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



A fucoidan fraction purified from Chnoospora minima; a potential inhibitor of LPS-induced inflammatory responses
Fernando et al.
International Journal of Biological Macromolecules, 104, 1185-1193 (2017)
Ewoud van Tricht et al.
Talanta, 166, 8-14 (2017-02-19)
During development of adenovirus-based vaccines, samples have to be analyzed in order to either monitor the production process or control the quality and safety of the product. An important quality attribute is the total concentration of intact adenoviruses, which currently
Danielle L Cullinane et al.
Reproduction (Cambridge, England), 149(1), 43-54 (2014-10-23)
The protamine 1 (Prm1) and sperm mitochondria-associated, cysteine-rich protein (Smcp) mRNAs exemplify a widespread pattern of mRNA-specific regulation of mRNA translation in post-meiotic spermatogenic cells, spermatids. Both mRNAs are transcribed and initially stored in free-mRNPs in early spermatids, and translated



Global Trade Item Number

SKUGTIN
D8906-500G04061835544240
D8906-5G04061835544264
D8906-100G04061835544233
D8906-50G04061835544257
D8906-10G04061835506262

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters