Skip to Content
Merck

Skip To

ABC299

Anti-FFAR2 Antibody/GPR43

from rabbit, purified by affinity chromatography

Synonym(s):

Free fatty acid receptor 2, G-protein coupled receptor 43, FFAR2/GPR43

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
13
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

species reactivity (predicted by homology)

human (based on 100% sequence homology), mouse

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FFAR2(2867)

General description

Free fatty acid receptor 2 (FFAR2), also known as G-protein coupled receptor 43 (GPR43), functions as the cell membrane receptor for short chain fatty acids that ultimately stimulates G protein coupled, inhibitory (Gi) signals via the adenylate cyclase pathway. FFAR2/GPR43 is found expressed in leukocytes in the lamina propria of the small intestine mostly but some expression can be also be found in splenocytes, adipose tissue, and enteroendocrine cells of the gut. Interesting, FFAR2/GPR43 because it senses short chain fatty acids which are often key metabolites of gut microbiota, it may serve as a biological sensor to regulate the activity of regulatory T cells and the immune response in diseases such as colitis.
~47 kDa observed. The calculated molecular weight of this protein is 37 kDa, but can be observed at ~47 kDa due to high glycosylation. Uncharacterized band(s) may appear in some lysates.

Immunogen

Epitope: Cytoplasmic domain
GST-tagged recombinant protein corresponding to the cytoplasmic domain of human FFAR2/GPR43.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-FFAR2 Antibody/GPR43 is validated for use in Western Blotting and Immunohistochemistry for the detection of FFAR2/GPR43.
Western Blotting Analysis: 1.0 µg/mL from a representative lot detected FFAR2/GPR43 in 10 µg of RAW264.7 cell lysate.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected FFAR2/GPR43 in mouse kidney and mouse liver tissue.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in CHEM-1 cell lysate.

Western Blotting Analysis: 1.0 µg/mL of this antibody detected FFAR2/GPR43 in 10 µg of CHEM-1 cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SAB1306868SAB4501283SAB2109184
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Gene Information

human ... FFAR2(2867)

Gene Information

human ... FFAR2(2867)

Gene Information

human ... FFAR2(2867)

Gene Information

human ... FFAR2(57396)

technique(s)

immunohistochemistry: suitable, western blot: suitable

technique(s)

western blot: 1:1000

technique(s)

ELISA: 1:1000, immunofluorescence: 1:100-1:500

technique(s)

western blot: 1 μg/mL


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Hideaki Fujiwara et al.
Nature communications, 9(1), 3674-3674 (2018-09-12)
Microbiome-derived metabolites influence intestinal homeostasis and regulate graft-versus-host disease (GVHD), but the molecular mechanisms remain unknown. Here we show the metabolite sensor G-protein-coupled receptor 43 (GPR43) is important for attenuation of gastrointestinal GVHD in multiple clinically relevant murine models. GPR43
Qian-Jiang Luo et al.
Gastroenterology report, 9(2), 154-165 (2021-05-25)
Butyrate acts as a regulator in multiple inflammatory organ injuries. However, the role of butyrate in acute liver injury has not yet been fully explored. In the present study, we aimed to investigate the association between butyrate and lipopolysaccharide (LPS)-induced
Yung-Mei Chao et al.
Nutrients, 14(20) (2022-10-28)
The role of short-chain fatty acids (SCFAs) in the brain on the developmental programming of hypertension is poorly understood. The present study explored dysregulated tissue levels of SCFAs and expression of SCFA-sensing receptors in the hypothalamic paraventricular nucleus (PVN), a



Global Trade Item Number

SKUGTIN
ABC29904053252985188

Questions

Reviews

No rating value

Active Filters