Skip to Content
Merck

Skip To

8.02412

2-Chloroacetamide

for synthesis

Synonym(s):

2-Chloroacetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C2H4ClNO
CAS Number:
Molecular Weight:
93.51
UNSPSC Code:
12352111
EC Index Number:
201-174-2
NACRES:
NA.22
MDL number:
Assay:
≥97.0% (GC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.0026 hPa ( 20 °C)

Quality Level

assay

≥97.0% (GC)

form

powder

potency

138 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rat)

pH

3.8-4.1 (20 °C, 40 g/L in H2O)

mp

115-118 °C

transition temp

flash point 170 °C

solubility

52.5 g/L

bulk density

760 kg/m3

storage temp.

2-30°C

SMILES string

ClCC(=[O+H])[N-H]

InChI

1S/C2H4ClNO/c3-1-2(4)5/h1H2,(H2,4,5)

InChI key

VXIVSQZSERGHQP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 116 °C
Melting range (upper value): ≤ 120 °C
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.186728.20290C0267
assay

≥97.0% (GC)

assay

-

assay

≥96.5% (GC)

assay

≥98%

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

300

solubility

52.5 g/L

solubility

-

solubility

-

solubility

-

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

form

powder

form

crystals

form

liquid

form

powder

mp

115-118 °C

mp

96-99 °C

mp

-

mp

116-118 °C (lit.)


Still not finding the right product?

Explore all of our products under 2-Chloroacetamide


pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Repr. 2 - Skin Sens. 1

Storage Class

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 2

flash_point_f

338.0 °F

flash_point_c

170 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
802412902504022536774619
802412100004022536373300
802412010004022536373294

Questions

Reviews

No rating value

Active Filters