Skip to Content
Merck

Skip To

1672803

USP

Transplatin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

trans-Platinum(II)diammine dichloride, trans-Diamminedichloroplatinum(II), trans-Dichlorodiammineplatinum(II)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
Pt(NH3)2Cl2
CAS Number:
Molecular Weight:
300.05
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

cisplatin

manufacturer/tradename

USP

mp

340 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N.N.Cl[Pt]Cl

InChI

1S/2ClH.2H3N.Pt/h2*1H;2*1H3;/q;;;;+2/p-2

InChI key

LXZZYRPGZAFOLE-UHFFFAOYSA-L

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BP544PHR32521134357
grade

pharmaceutical primary standard

grade

-

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

BP

manufacturer/tradename

-

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

pharmaceutical (small molecule)

format

neat

format

-

format

-

format

neat

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-30°C

storage temp.

-

mp

340 °C (dec.) (lit.)

mp

340 °C (dec.) (lit.)

mp

340 °C (dec.) (lit.)

mp

270 °C (lit.)


Still not finding the right product?

Explore all of our products under Transplatin


pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



James Chih-Hsin Yang et al.
The Lancet. Oncology, 16(2), 141-151 (2015-01-16)
We aimed to assess the effect of afatinib on overall survival of patients with EGFR mutation-positive lung adenocarcinoma through an analysis of data from two open-label, randomised, phase 3 trials. Previously untreated patients with EGFR mutation-positive stage IIIB or IV
Wei Qian et al.
Oncotarget, 5(12), 4180-4194 (2014-06-24)
Overcoming platinum drug resistance represents a major clinical challenge in cancer treatment. We discovered a novel drug combination using cisplatin and a class of thioquinazolinone derivatives including mdivi-1 (mitochondrial division inhibitor-1), that induces synergistic apoptosis in platinum resistant tumor cells
Yuniardini S Wimardhani et al.
Journal of oral science, 56(2), 119-126 (2014-06-17)
Chitosan, a multipurpose biomaterial, has been shown to exert effects against several types of cancer including oral cancer. However, the mechanisms underlying the anticancer activities of chitosan on oral squamous cell carcinoma (SCC) cells remain largely unknown. The present study



Global Trade Item Number

SKUGTIN
1672803-25MG04061833833988

Questions

Reviews

No rating value

Active Filters