Skip to Content
Merck

Skip To

1592409

USP

Quercetin

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Quercetin dihydrate, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-1-benzopyran-4-one dihydrate, 3,3′,4′,5,7-Pentahydroxyflavone dihydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H10O7 · 2H2O
CAS Number:
Molecular Weight:
338.27
MDL number:
NACRES:
NA.24
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
317313
Colour Index Number:
75670
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

quercetin

manufacturer/tradename

USP

mp

>300 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

OC(C(O)=C1)=CC=C1C2=C(O)C(C3=C(O)C=C(O)C=C3O2)=O

InChI

1S/C15H10O7.2H2O/c16-7-4-10(19)12-11(5-7)22-15(14(21)13(12)20)6-1-2-8(17)9(18)3-6;;/h1-5,16-19,21H;2*1H2

InChI key

GMGIWEZSKCNYSW-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Quercetin USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Cranberry Fruit Juice Dry Extract
  • Japanese Sophora Flower
  • Japanese Sophora Flower Powder
  • Japanese Sophora Flower Bud
  • Japanese Sophora Flower Dry Extract
  • Cranberry Fruit Juice Dry Extract Capsules
  • Rutin

Biochem/physiol Actions

Quercetin dihydrate is a flavonoid with anticancer activity.
Quercetin dihydrate is a flavonoid with anticancer activity. Quercetin is a mitochondrial ATPase and phosphodiesterase inhibitor. It Inhibits PI3-kinase activity and slightly inhibits PIP kinase activity. Quercetin has antiproliferative effects on cancer cell lines, reduces cancer cell growth via type II estrogen receptors, and arrests human leukemic T cells in late G1 phase of the cell cycle.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHL89262Q4951PHR1488
grade

pharmaceutical primary standard

grade

primary reference standard

grade

-

grade

certified reference material, pharmaceutical secondary standard

manufacturer/tradename

USP

manufacturer/tradename

PhytoLab

manufacturer/tradename

-

manufacturer/tradename

-

format

neat

format

neat

format

-

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

food and beverages

application(s)

-

application(s)

pharmaceutical (small molecule)

mp

>300 °C (lit.)

mp

-

mp

316.5 °C

mp

-

storage temp.

−20°C

storage temp.

-

storage temp.

room temp

storage temp.

-10 to -25°C


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Catarina Cardoso et al.
PloS one, 9(8), e104201-e104201 (2014-08-16)
Strigolactones (SLs) trigger germination of parasitic plant seeds and hyphal branching of symbiotic arbuscular mycorrhizal (AM) fungi. There is extensive structural variation in SLs and plants usually produce blends of different SLs. The structural variation among natural SLs has been
Gabriella Testa et al.
PloS one, 9(5), e96795-e96795 (2014-05-08)
Chronic inflammatory events appear to play a fundamental role in Alzheimer's disease (AD)-related neuropathological changes, and to result in neuronal dysfunction and death. The inflammatory responses observed in the AD brain include activation and proliferation of glial cells, together with
Guozheng Huang et al.
European journal of medicinal chemistry, 81, 15-21 (2014-05-14)
Two sets of carbamates based on the natural alkaloid evodiamine were designed, synthesized and evaluated as potential butyrylcholinesterase inhibitors. Although a set of carbamates of 3-hydroxyevodiamine (10a-f) is inactive both at AChE and BChE, carbamates of 5-deoxo-3-hydroxyevodiamine (11a-f) exhibit much



Global Trade Item Number

SKUGTIN
1592409-500MG04061838744463
1592409-200MG04061841203940

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is the product Quercetin (1592409) is soluble in water or DMSO?

    1 answer
    1. Solubility information for this item is not provided. As it is a USP reference standard, inquiries about it should be directed to USP.

      Helpful?

Reviews

No rating value

Active Filters