Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
neomycin
form
solid
packaging
pkg of 150 mg
manufacturer/tradename
USP
application(s)
USP Biologics
pharmaceutical
format
neat
storage temp.
−20°C
SMILES string
[S](=O)(=O)(O)O.N[C@H]1[C@H](O[C@H]([C@H]([C@@H]1O)O)CN)O[C@@H]2[C@H](O[C@H]([C@@H]2O)O[C@H]3[C@@H]([C@H](C[C@H]([C@@H]3O)N)N)O[C@H]4O[C@@H]([C@H]([C@@H]([C@H]4N)O)O)CN)CO
InChI
1S/C23H46N6O13.H2O4S/c24-2-7-13(32)15(34)10(28)21(37-7)40-18-6(27)1-5(26)12(31)20(18)42-23-17(36)19(9(4-30)39-23)41-22-11(29)16(35)14(33)8(3-25)38-22;1-5(2,3)4/h5-23,30-36H,1-4,24-29H2;(H2,1,2,3,4)/t5-,6+,7-,8+,9-,10-,11-,12+,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23+;/m1./s1
InChI key
OIXVKQDWLFHVGR-WQDIDPJDSA-N
General description
Find answers to frequently asked questions on USP products in USP FAQs.
Application
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| packaging pkg of 150 mg | packaging - | packaging pkg of 20 mg | packaging pkg of 100 mg |
| storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C | storage temp. 2-8°C |
| API family neomycin | API family neomycin | API family nicardipine | API family decitabine |
| format neat | format neat | format neat | format neat |
| application(s) USP Biologics | application(s) USP Biologics | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| form solid | form solid | form - | form - |
signalword
Warning
hcodes
Hazard Classifications
Skin Sens. 1
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

