Skip to Content
Merck

Skip To

1317007

USP

Hydrocortisone acetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Hydrocortisone 21-acetate, 11β,17α,21-Trihydroxy-4-pregnene-3,20-dione 21-acetate, 17-α-Hydroxycorticosterone acetate, 17-Hydroxycorticosterone 21-acetate, 21-Acetoxy-4-pregnene-11β,17α-diol-3,20-dione, 4-Pregnene-11β,17α,21-triol-3,20-dione 21-acetate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C23H32O6
CAS Number:
Molecular Weight:
404.50
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
2066841
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

hydrocortisone

manufacturer/tradename

USP

mp

223 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])[C@@H](O)C[C@@]4(C)[C@@]2([H])CC[C@]4(O)C(=O)COC(C)=O

InChI

1S/C23H32O6/c1-13(24)29-12-19(27)23(28)9-7-17-16-5-4-14-10-15(25)6-8-21(14,2)20(16)18(26)11-22(17,23)3/h10,16-18,20,26,28H,4-9,11-12H2,1-3H3/t16-,17-,18-,20+,21-,22-,23-/m0/s1

InChI key

ALEXXDVDDISNDU-JZYPGELDSA-N

Gene Information

human ... NR3C1(2908)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hydrocortisone acetate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Hydrocortisone Acetate Cream
  • Hydrocortisone Acetate Lotion
  • Hydrocortisone Acetate Ointment
  • Neomycin Sulfate and Hydrocortisone Acetate Ointment
  • Neomycin Sulfate and Hydrocortisone Acetate Ophthalmic Suspension
  • Oxytetracycline Hydrochloride and Hydrocortisone Acetate Ophthalmic Suspension
  • Neomycin and Polymyxin B Sulfates and Hydrocortisone Acetate Cream

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1606Y0001482H1400000
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

EDQM

manufacturer/tradename

EDQM

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

mp

223 °C (dec.) (lit.)

mp

223 °C (dec.) (lit.)

mp

223 °C (dec.) (lit.)

mp

223 °C (dec.) (lit.)

Gene Information

human ... NR3C1(2908)

Gene Information

human ... NR3C1(2908)

Gene Information

human ... NR3C1(2908)

Gene Information

human ... NR3C1(2908)


Still not finding the right product?

Explore all of our products under Hydrocortisone acetate


pictograms

Health hazard

signalword

Danger

Hazard Classifications

Repr. 1B - STOT RE 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Adamo Fini et al.
AAPS PharmSciTech, 9(3), 762-768 (2008-06-20)
The purpose of this research was the preparation of four formulations containing hydrocortisone acetate (HCA) for topical application, including two aqueous systems (hydrophilic microemulsion and aqueous gel) and two systems with dominant hydrophobicity (hydrophobic microemulsion and ointment). The formulations were
Janyce A Sugui et al.
Journal of clinical microbiology, 52(10), 3707-3721 (2014-08-08)
Aspergillus section Fumigati contains 12 clinically relevant species. Among these Aspergillus species, A. fumigatus is the most frequent agent of invasive aspergillosis, followed by A. lentulus and A. viridinutans. Genealogical concordance and mating experiments were performed to examine the relationship
Timothy Kassis et al.
PLoS neglected tropical diseases, 8(11), e3305-e3305 (2014-11-21)
Lymphatic Filariasis, a Neglected Tropical Disease, is caused by thread-like parasitic worms, including B. malayi, which migrate to the human lymphatic system following transmission. The parasites reside in collecting lymphatic vessels and lymph nodes for years, often resulting in lymphedema



Global Trade Item Number

SKUGTIN
1317007-200MG04061838728531

Questions

Reviews

No rating value

Active Filters