Skip to Content
Merck

Skip To

1294987

USP

L-Glutamic acid hydrochloride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(S)-2-Aminoglutaric acid, (S)-2-Aminopentanedioic acid ammonium salt, Glu HCl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C5H9NO4 · HCl
CAS Number:
Molecular Weight:
183.59
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3565569
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

glutamic acid

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cl.N[C@@H](CCC(O)=O)C(O)=O

InChI

1S/C5H9NO4.ClH/c6-3(5(9)10)1-2-4(7)8;/h3H,1-2,6H2,(H,7,8)(H,9,10);1H/t3-;/m0./s1

InChI key

RPAJSBKBKSSMLJ-DFWYDOINSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

ʟ-Glutamic acid hydrochloride USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Biochem/physiol Actions

Agonist at kainate, NMDA, and quisqualate receptors; an excitatory amino acid neurotransmitter.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
G21288.146321012517
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

USP

grade

pharmaceutical primary standard

grade

-

grade

-

grade

pharmaceutical primary standard

format

neat

format

-

format

-

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

application(s)

pharmaceutical (small molecule)

API family

glutamic acid

API family

-

API family

-

API family

glutamine


Still not finding the right product?

Explore all of our products under L-Glutamic acid hydrochloride


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1294987-100MG04061833830475

Questions

Reviews

No rating value

Active Filters