Skip to Content
Merck

Skip To

1270402

USP

Finasteride

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

N-(2-methyl-2-propyl)-3-oxo-4-aza-5α-androst-1-ene-17β-carboxamide, N-tert-Butyl-3-oxo-4-aza-5α-androst-1-en-17β-carboxamide, MK-906

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C23H36N2O2
CAS Number:
Molecular Weight:
372.54
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

finasteride

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12CC[C@@]3([H])[C@]4([H])CC[C@H](C(=O)NC(C)(C)C)[C@@]4(C)CC[C@]3([H])[C@@]1(C)C=CC(=O)N2

InChI

1S/C23H36N2O2/c1-21(2,3)25-20(27)17-8-7-15-14-6-9-18-23(5,13-11-19(26)24-18)16(14)10-12-22(15,17)4/h11,13-18H,6-10,12H2,1-5H3,(H,24,26)(H,25,27)/t14-,15-,16-,17+,18+,22-,23+/m0/s1

InChI key

DBEPLOCGEIEOCV-WSBQPABSSA-N

Gene Information

human ... SRD5A2(6716)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Finasteride USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Finasteride Tablets

Biochem/physiol Actions

Selective 5α-reductase inhibitor; antiandrogen.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0000090BP740Y0001604
manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

BP

manufacturer/tradename

EDQM

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

API family

finasteride

API family

finasteride

API family

finasteride

API family

finasteride

Gene Information

human ... SRD5A2(6716)

Gene Information

human ... SRD5A2(6716)

Gene Information

human ... SRD5A2(6716)

Gene Information

human ... SRD5A2(6716)


Still not finding the right product?

Explore all of our products under Finasteride


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1270402-200MG04061838724670

Questions

Reviews

No rating value

Active Filters