Skip to Content
Merck

Skip To

1269403

USP

Fenbendazole

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Methyl 5-(phenylthio)-2-benzimidazolecarbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C15H13N3O2S
CAS Number:
Molecular Weight:
299.35
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
759077
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

fenbendazole

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

COC(=O)Nc1nc2cc(Sc3ccccc3)ccc2[nH]1

InChI

1S/C15H13N3O2S/c1-20-15(19)18-14-16-12-8-7-11(9-13(12)17-14)21-10-5-3-2-4-6-10/h2-9H,1H3,(H2,16,17,18,19)

InChI key

HDDSHPAODJUKPD-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Fenbendazole USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Oxfendazole

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
F0040000BP69235032
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

analytical standard

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

BP

manufacturer/tradename

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

clinical testing

format

neat

format

neat

format

neat

format

neat

API family

fenbendazole

API family

fenbendazole

API family

fenbendazole

API family

-


Still not finding the right product?

Explore all of our products under Fenbendazole


pictograms

Health hazardEnvironment

signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2 - STOT RE 2

target_organs

Liver,lymph node,Stomach,Nervous system

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Vanessa O Ezenwa et al.
Science (New York, N.Y.), 347(6218), 175-177 (2015-01-13)
Parasitic worms modulate host immune responses in ways that affect microbial co-infections. For this reason, anthelmintic therapy may be a potent tool for indirectly controlling microbial pathogens. However, the population-level consequences of this type of intervention on co-infecting microbes are
David Villar et al.
Journal of the American Association for Laboratory Animal Science : JAALAS, 46(6), 8-15 (2007-11-13)
This review summarizes findings from toxicologic, carcinogenic, immunologic, and metabolic studies on fenbendazole (FBZ). Currently, FBZ is used to treat or prevent pinworm outbreaks in laboratory rodents. Because antiparasitic treatments usually are not part of experimental designs, interactions from the
Bettencourt P S Capece et al.
Veterinary journal (London, England : 1997), 181(3), 241-250 (2009-01-07)
Albendazole and fenbendazole are methylcarbamate benzimidazole anthelmintics extensively used to control gastrointestinal parasites in domestic animals. These parent compounds are metabolised to albendazole sulfoxide and fenbendazole sulfoxide (oxfendazole), respectively. Both sulfoxide derivatives are anthelmintically active and are manufactured for use



Global Trade Item Number

SKUGTIN
1269403-100MG04061833020043

Questions

Reviews

No rating value

Active Filters