Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
pharmaceutical primary standard
agency
USP
API family
felbamate
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
NC(=O)OCC(COC(N)=O)c1ccccc1
InChI
1S/C11H14N2O4/c12-10(14)16-6-9(7-17-11(13)15)8-4-2-1-3-5-8/h1-5,9H,6-7H2,(H2,12,14)(H2,13,15)
InChI key
WKGXYQFOCVYPAC-UHFFFAOYSA-N
Gene Information
human ... GRIN1(2902), GRIN2A(2903), GRIN2B(2904), GRIN2C(2905), GRIN2D(2906), GRIN3A(116443), GRIN3B(116444)
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Also, for use with USP monographs such as:
- Felbamate Tablets
- Felbamate Oral Suspension
Analysis Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename USP | manufacturer/tradename - | manufacturer/tradename USP | manufacturer/tradename USP |
| grade pharmaceutical primary standard | grade - | grade pharmaceutical primary standard | grade pharmaceutical primary standard |
| format neat | format - | format neat | format neat |
| application(s) pharmaceutical (small molecule) | application(s) - | application(s) pharmaceutical (small molecule) | application(s) pharmaceutical (small molecule) |
| API family felbamate | API family - | API family felbamate | API family felbamate |
| Gene Information human ... GRIN1(2902), GRIN2A(2903), GRIN2B(2904), GRIN2C(2905), GRIN2D(2906), GRIN3A(116443), GRIN3B(116444) | Gene Information human ... GRIN1(2902), GRIN2A(2903), GRIN2B(2904), GRIN2C(2905), GRIN2D(2906), GRIN3A(116443), GRIN3B(116444) | Gene Information - | Gene Information - |
Still not finding the right product?
Explore all of our products under Felbamate
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

