Skip to Content
Merck

Skip To

1096826

USP

Castor oil

United States Pharmacopeia (USP) Reference Standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

castor oil

manufacturer/tradename

USP

refractive index

n20/D 1.478 (lit.)

bp

313 °C (lit.)

application(s)

pharmaceutical

SMILES string

O(C(COC(=O)CCCCCCC\C=C/CC(O)CCCCCC)COC(=O)CCCCCCC\C=C/CC(O)CCCCCC)C(=O)CCCCCCC\C=C/CC(O)CCCCCC

InChI

1S/C57H104O9/c1-4-7-10-31-40-51(58)43-34-25-19-13-16-22-28-37-46-55(61)64-49-54(66-57(63)48-39-30-24-18-15-21-27-36-45-53(60)42-33-12-9-6-3)50-65-56(62)47-38-29-23-17-14-20-26-35-44-52(59)41-32-11-8-5-2/h25-27,34-36,51-54,58-60H,4-24,28-33,37-50H2,1-3H3/b

InChI key

ZEMPKEQAKRGZGQ-AAKVHIHISA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Castor oil USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR2972C960683912
manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

-

application(s)

pharmaceutical

application(s)

pharmaceutical

application(s)

-

application(s)

-

bp

313 °C (lit.)

bp

313 °C (lit.)

bp

313 °C (lit.)

bp

313 °C (lit.)

API family

castor oil

API family

castor oil

API family

-

API family

-

refractive index

n20/D 1.478 (lit.)

refractive index

n20/D 1.478 (lit.)

refractive index

n20/D 1.478 (lit.)

refractive index

n20/D 1.478 (lit.)


Still not finding the right product?

Explore all of our products under Castor oil


Storage Class

10 - Combustible liquids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters