Skip to Content
Merck

Skip To

1095506

USP

Carbidopa

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropionic acid monohydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H14N2O4 · H2O
CAS Number:
Molecular Weight:
244.24
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

API family

carbidopa

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.C[C@@](Cc1ccc(O)c(O)c1)(NN)C(O)=O

InChI

1S/C10H14N2O4.H2O/c1-10(12-11,9(15)16)5-6-2-3-7(13)8(14)4-6;/h2-4,12-14H,5,11H2,1H3,(H,15,16);1H2/t10-;/m0./s1

InChI key

QTAOMKOIBXZKND-PPHPATTJSA-N

Gene Information

human ... DDC(1644)

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Carbidopa USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Carbidopa and Levodopa Extended-Release Tablets
  • Carbidopa and Levodopa Orally Disintegrating Tablets
  • Carbidopa and Levodopa Tablets

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BP060C0460000PHR1655
Carbidopa British Pharmacopoeia (BP) Reference Standard

BP060

Carbidopa

manufacturer/tradename

USP

manufacturer/tradename

BP

manufacturer/tradename

EDQM

manufacturer/tradename

-

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

format

neat

format

neat

format

neat

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical
pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

API family

carbidopa

API family

carbidopa

API family

carbidopa

API family

carbidopa

Gene Information

human ... DDC(1644)

Gene Information

human ... DDC(1644)

Gene Information

human ... DDC(1644)

Gene Information

human ... DDC(1644)


Still not finding the right product?

Explore all of our products under Carbidopa


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Q. Alan Xu, Timothy L. Madden
Analytical Methods for Therapeutic Drug Monitoring and Toxicology, 80-80 (2011)
Mohseni Mehran S M et al.
Journal of clinical and diagnostic research : JCDR, 7(6), 1004-1007 (2013-08-02)
According to many studies, sprouted fava beans are a rich source of levo-dihydroxy phenylalanine (L-dopa) the precursor of dopamine, and they are now being investigated for use in the management of Parkinson's disease. The addition of Carbidopa (C-dopa) can reduce
A Castro et al.
Neurologia (Barcelona, Spain), 20(4), 180-188 (2005-05-14)
Levodopa remains the mainstay treatment for Parkinson's disease (PD). Chronic treatment is associated with motor complications (MC) that marred the clinical benefit of levodopa. These problems and experimental data in cell cultures indicating a neurotoxic effect of levodopa have led



Global Trade Item Number

SKUGTIN
1095506-400MG04061838688651

Questions

Reviews

No rating value

Active Filters