Skip to Content
Merck

Skip To

1012123

USP

Adenosine

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

9-β-D-Ribofuranosyladenine, Adenine riboside, Adenine-9-β-D-ribofuranoside

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H13N5O4
CAS Number:
Molecular Weight:
267.24
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
93029
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

description

API family: adenosine

API family

adenosine

form

solid

manufacturer/tradename

USP

mp

234-236 °C (lit.)

application(s)

USP Biologics
pharmaceutical (small molecule)

format

neat

SMILES string

Nc1ncnc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O

InChI

1S/C10H13N5O4/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(18)6(17)4(1-16)19-10/h2-4,6-7,10,16-18H,1H2,(H2,11,12,13)/t4-,6-,7-,10-/m1/s1

InChI key

OIRDTQYFTABQOQ-KQYNXXCUSA-N

Gene Information

General description

Adenosine is a nucleoside formed by the attachment of adenine to a ribose sugar. The USP Adenosine standard provides a reference for the quality and purity of adenosine used in pharmaceutical formulations and research applications, ensuring consistency in its therapeutic and experimental use.

The USP biologics oligonucleotides category includes a variety of synthetic nucleic acid sequences that are essential in research, diagnostics, and therapeutic applications. These oligonucleotides play crucial roles in molecular biology, including gene expression regulation, antisense therapy, and RNA interference. The USP provides comprehensive standards, reference materials, and analytical procedures to ensure the identity, quality, purity, and consistency of oligonucleotide products throughout their lifecycle.

The United States Pharmacopeia (USP) provides quality standards for biologics to ensure their safety, efficacy, and quality throughout the manufacturing process. These standards assist manufacturers in adhering to regulatory requirements and help safeguard public health by reducing risks associated with biologics.

Application

Adenosine USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.
Also, for use with USP monographs such as:
  • Adenosine Injection
  • Heparin Sodium

Biochem/physiol Actions

Endogenous neurotransmitter at adenosine receptors. Cardioprotective effects may relate to activation of A1 adenosine receptors. The antiplatelet and anti−inflammatory actions of adenosine appear to be mediated via the A2 adenosine receptor. In contrast, adenosine appears to be a pro-inflammatory mediator in asthma and chronic obstructive pulmonary disease (COPD).
Endogenous neurotransmitter. Cardioprotective effects may relate to activation of A1 adenosine receptors. The antiplatelet and anti−inflammatory actions of adenosine appear to be mediated via the A2 adenosine receptor. In contrast, adenosine appears to be a pro-inflammatory mediator in asthma and chronic obstructive pulmonary disease (COPD).

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
This product is part of USP Biologics Program.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHL8097701890A0230200
grade

pharmaceutical primary standard

grade

primary reference standard

grade

-

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

PhytoLab

manufacturer/tradename

-

manufacturer/tradename

EDQM

form

solid

form

solid

form

powder

form

-

format

neat

format

-

format

-

format

neat

application(s)

USP Biologics
pharmaceutical (small molecule)

application(s)

-

application(s)

-

application(s)

pharmaceutical (small molecule)

Gene Information

human ... ADORA1(134), ADORA2A(135), ADORA2B(136), ADORA3(140)

Gene Information

human ... ADORA1(134), ADORA2A(135), ADORA2B(136), ADORA3(140)

Gene Information

human ... ADORA1(134), ADORA2A(135), ADORA2B(136), ADORA3(140)

Gene Information

human ... ADORA1(134), ADORA2A(135), ADORA2B(136), ADORA3(140)


Still not finding the right product?

Explore all of our products under Adenosine


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
1012123-200MG04061838667182

Questions

Reviews

No rating value

Active Filters