Skip to Content
Merck

Skip To

V5625

Snake venom from Bothrops jararaca

Factor X activator, lyophilized

Synonym(s):

South American Pit Viper

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
Form:
lyophilized
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Snake venom from Bothrops jararaca,

form

lyophilized

Quality Segment

storage temp.

2-8°C

Biochem/physiol Actions

Snake venom from Bothrops jararaca mainly contains three types of toxins snake venom metalloproteinases (SVMP), snake venom serine proteases (SVSP), and C-type lectin-like proteins (CTLP). These toxins aids in the pathogenesis of envenomation by activating blood coagulation prothrombin, von Willebrand factor (VWF), and factor X, and by altering the function of protein C, kininogen, and blood platelets.[1]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
V82501509019SAB4700938
form

lyophilized

form

-

form

-

form

buffered aqueous solution

Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

-

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

15-25°C

storage temp.

2-8°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
V5625-100MG04061832943558
V5625-1G04061826694800

Questions

  1. What is the composition of V5625 Snake venom from Bothrops jararaca?

    What is the composition of V5625 Snake venom from Bothrops jararaca?

    1 answer
    1. Product V5625 (Snake venom from Bothrops jararaca) is a crude extract. It contains a number of compounds, including carbohydrates and proteins. The individual components have not been identified. Please see the link below to a publication that may be helpful.
      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC8402457/#:~:text=These%20data%20suggest%20that%20sub,cancer%20cell%20growth%20and%20survival.

      Helpful?

Reviews

No rating value

Active Filters