Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
CAS Number:
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12161900
MDL number:
Pricing and availability is not currently available.
Quality Level
CMC
0.018 mM
transition temp
cloud point 94.3 °C
solubility
water: miscible
SMILES string
[H]C([H])=O.OCCO.CC(C)(C)CC(C)(C)c1ccc(O)cc1
InChI
1S/C14H22O.C2H6O2.CH2O/c1-13(2,3)10-14(4,5)11-6-8-12(15)9-7-11;3-1-2-4;1-2/h6-9,15H,10H2,1-5H3;3-4H,1-2H2;1H2
InChI key
GWJOFBXSBDVUMH-UHFFFAOYSA-N
General description
A nonionic liquid polymer of the alkyl aryl polyether alcohol type. Used as a surfactant.
Application
Tyloxapol is a nonionic liquid polymer of the alkyl aryl polyether alcohol type. It is used as a surfactant to aid liquefaction. Tyloxapol is used in the development of non-ionic surfactant-based liposomes (niosomes) for use in drug delivery systems. Tyloxapol may be used to study its mechanism of protecting macrophages from endotoxins. Tyloxapol may be used to study is potential cell toxicitiy.
1 of 1
This Item | |||
|---|---|---|---|
| solubility water: miscible | solubility - | solubility - | solubility - |
| Quality Level 200 | Quality Level 100 | Quality Level 200 | Quality Level - |
| CMC 0.018 mM | CMC 0.018 mM | CMC - | CMC 0.018 mM |
| transition temp cloud point 94.3 °C | transition temp cloud point 94.3 °C | transition temp - | transition temp cloud point 94.3 °C |
Still not finding the right product?
Explore all of our products under Tyloxapol
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Jung-hua Steven Kuo et al.
Pharmaceutical research, 23(7), 1509-1516 (2006-06-17)
Tyloxapol, a viscous polymer of the alkyl aryl polyether alcohol type, is classified as a nonionic surfactant and is widely used in biomedical applications. Although tyloxapol has been reported to be cytotoxic in various cell lines, there is no published
Jung-Hua Steven Kuo et al.
Colloids and surfaces. B, Biointerfaces, 64(2), 208-215 (2008-03-14)
Tyloxapol is reported to prevent macrophages from reacting to endotoxin. However, the intracellular responses that tyloxapol induces in macrophages are still not fully explored. Hence, the objective of this study was to evaluate the intracellular events in macrophages treated with
Tariq Al-Qirim et al.
Archiv der Pharmazie, 345(5), 401-406 (2012-01-24)
A new series of N-(benzoylphenyl) and N-(acetylphenyl)-1-benzofuran-2-carboxamides (3a-3d and 4a'-4c') were synthesized. Compounds (3a, 3b, and 4a'-4c') were tested in vivo using Triton-WR-1339-induced hyperlipidemic rats as an experimental model for their hypolipidemic activity. The tested animals were divided into eight
Global Trade Item Number
| SKU | GTIN |
|---|---|
| T8761-50G | 04061835550340 |


