Skip to Content
Merck

Skip To

T7004

Thymidine 5′-monophosphate disodium salt hydrate

≥99%

Synonym(s):

5′-Thymidylic acid disodium salt, TMP, dTMP

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H13N2Na2O8P · xH2O
CAS Number:
Molecular Weight:
366.17 (anhydrous basis)
UNSPSC Code:
41106305
NACRES:
NA.51
PubChem Substance ID:
EC Number:
251-518-0
Beilstein/REAXYS Number:
4086927
MDL number:
Assay:
≥99%
Biological source:
synthetic (organic)
Form:
powder
Solubility:
H2O: 100 mg/mL to clear, colorless
Storage temp.:
−20°C
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic (organic)

Quality Segment

assay

≥99%

form

powder

solubility

H2O: 100 mg/mL to clear, colorless

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].CC1=CN([C@H]2C[C@H](O)[C@@H](COP([O-])([O-])=O)O2)C(=O)NC1=O

InChI

1S/C10H15N2O8P.2Na.H2O/c1-5-3-12(10(15)11-9(5)14)8-2-6(13)7(20-8)4-19-21(16,17)18;;;/h3,6-8,13H,2,4H2,1H3,(H,11,14,15)(H2,16,17,18);;;1H2/q;2*+1;/p-2/t6-,7+,8+;;;/m0.../s1

InChI key

KCOPAVKVHIWPKB-SPSULGLQSA-L

General description

Thymidine 5′-monophosphate de novo synthesis from deoxyuridine monophosphate (dUMP) is catalyzed by the enzyme thymidylate synthase.

Application

Thymidine 5′-monophosphate disodium salt hydrate has been used as a standard for the quantification of metabolites from plasma and tumor interstitial fluid using liquid chromatography-mass spectrometry analysis (LC/MS). It has also been used as a single deoxy-mononucleotide to test its effect on the cell growth of human malignant intestinal CaCo-2 cells and normal rat intestinal cell line IEC-6 cells.[1]
Thymidine 5′-monophosphate disodium salt hydrate has been used as a standard solution.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
U6375D3876T4510
biological source

synthetic (organic)

biological source

(chemical)

biological source

synthetic (organic)

biological source

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

powder

assay

≥99%

assay

≥99%

assay

≥98% (HPLC)

assay

≥98% (HPLC)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

solubility

H2O: 100 mg/mL to clear, colorless

solubility

water: 50 mg/mL, clear, colorless

solubility

water: 50 mg/mL, clear, colorless

solubility

water: 50 mg/mL


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
T7004-1G04061837372186
T7004-250MG04061833265772
T7004-100MG04061833265765

Questions

Reviews

No rating value

Active Filters