Skip to Content
Merck

Skip To

T6791

Trizma® base

BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)

Synonym(s):

2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Tris(hydroxymethyl)aminomethane, Trometamol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
UNSPSC Code:
12352104
NACRES:
NA.25
PubChem Substance ID:
EC Number:
201-064-4
Beilstein/REAXYS Number:
741883
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

aminopeptidase substrate

Quality Segment

product line

BioXtra

assay

≥99.9% (titration)

form

crystalline

storage condition

dry at room temperature

impurities

≤0.005% Insoluble matter

ign. residue (900°C)

≤0.01% (as SO4)

loss

≤0.5% loss on drying, 110°C

color

white

pH

10.5-12.0 (1 M in H2O)

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)

mp

167-172 °C (lit.)

solubility

H2O: 1 M, colorless, water: 678 g/L at 20 °C

anion traces

chloride (Cl-): ≤0.005%, sulfate (SO42-): ≤0.005%

cation traces

Al: ≤0.0005%, As: ≤0.0001%, Ba: ≤0.0005%, Bi: ≤0.0005%, Ca: ≤0.001%, Cd: ≤0.0005%, Co: ≤0.0005%, Cr: ≤0.0005%, Cu: ≤0.0005%, Fe: ≤0.0005%, K: ≤0.005%, Li: ≤0.0005%, Mg: ≤0.0005%, Mn: ≤0.0005%, Mo: ≤0.0005%, Na: ≤0.005%, Ni: ≤0.0005%, Pb: ≤0.0005%, Sr: ≤0.0005%, Zn: ≤0.0005%

SMILES string

NC(CO)(CO)CO

absorption

≤0.020 at 280 in H2O at 1 M, ≤0.025 at 260 in H2O at 1 M

application(s)

diagnostic assay manufacturing
life science and biopharma
sample preparation

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

General description

Tris base, also referred as THAM or Trizma® base, plays a vital role in various research applications as a biological buffer. It is commonly employed in buffer formulations like TAE and TBE buffers, thanks to its pKa of 8.06, which makes it suitable for maintaining pH within the physiological range (pH 7.0-9.0) for most living organisms. However, it is important to exercise caution when studying proteins, as Tris may interfere with the activity of certain enzymes. This versatile compound is indispensable in laboratories, where it is utilized to prepare buffers compatible with biological fluids, serve as a standard pH solution, and facilitate procedures such as lactate dehydrogenase assays, in situ hybridization, and protein extraction from cells. Moreover, Tris base finds applications in biochemistry and molecular biology labs aiding in cell membrane permeability studies and buffer preparation.

Application

Tris buffer has been used to stabilize the pH, facilitating the measurement of cytotoxicity using the sulforhodamine B (SRB) colorimetric cytotoxicity assay in cancer research. Additionally, it was utilized in this study to regulate the pH of the incubation medium to assess the glucose secretion as a response mediated by glucocorticoids in metabolic pathways.

Features and Benefits

  • Suitable for Cell Membrane permeability studies and for buffer preparation in Biochemical and Molecular Biology Research
  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications
  • Effective Buffering from pH 7-9 (25 °C) with a pKa of 8.1 (25 °C)
  • Tested for the presence of Anionic and Cationic traces

Other Notes

For additional information on our range of Biochemicals, please complete this form.
The pH values of all buffers are temperature- and concentration-dependent. For Tris buffers, pH increases about 0.03 unit per °C decrease in temperature, and decreases 0.03-0.05 unit per ten-fold dilution.
For precise applications, use a carefully calibrated pH meter with a glass/calomel combination electrode.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T606693362RDD008
assay

≥99.9% (titration)

assay

≥99.9% (titration)

assay

≥99.8% (T)

assay

≥99.9%

product line

BioXtra

product line

-

product line

BioUltra

product line

Redi-Dri

Quality Level

400

Quality Level

400

Quality Level

200

Quality Level

100

application(s)

diagnostic assay manufacturing
life science and biopharma
sample preparation

application(s)

clinical research
clinical testing
diagnostic assay manufacturing

application(s)

-

application(s)

-

form

crystalline

form

crystalline powder

form

crystalline

form

powder

pH

10.5-12.0 (1 M in H2O)

pH

10.0-11.5

pH

10.5-12.0 (25 °C, 4 M in H2O)

pH

10.5-12, 7-9


Still not finding the right product?

Explore all of our products under Trizma® base


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters