Skip to Content
Merck

Skip To

T6394

Timolol maleate salt

≥98% (TLC), β-adrenoceptor antagonist, powder

Synonym(s):

S-(−)-1-(t-Butylamino)-3-[(4-morpholino-1,2,5-thiadiazol-3-yl)oxy]-2-propanol maleate salt, Timolol hydrogen maleate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H24N4O3S · C4H4O4
CAS Number:
Molecular Weight:
432.49
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
248-111-5
MDL number:
Assay:
≥98% (TLC)
Form:
powder
Quality level:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Timolol maleate salt, ≥98% (TLC), powder

Quality Level

assay

≥98% (TLC)

form

powder

optical activity

[α]25/D -5.4°, c = 4.9 in 1 M HCl(lit.)

color

white to off-white

mp

202-203 °C (lit.)

solubility

H2O: soluble, ethanol: soluble

ε (extinction coefficient)

8,700 at 294 nm at 1 M

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

SMILES string

[H]\C(=C(/[H])C(O)=O)C(O)=O.CC(C)(C)NC[C@H](O)COc1nsnc1N2CCOCC2

InChI

1S/C13H24N4O3S.C4H4O4/c1-13(2,3)14-8-10(18)9-20-12-11(15-21-16-12)17-4-6-19-7-5-17;5-3(6)1-2-4(7)8/h10,14,18H,4-9H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t10-;/m0./s1

InChI key

WLRMANUAADYWEA-NWASOUNVSA-N

Gene Information

Application

Timolol maleate has been used as an adrenergic β-blocker for evaluating cardiovascular functions in mice5. It has also been used for evaluating the uptake of 86Rb+ in rat cardiomyocytes6.

Biochem/physiol Actions

Timolol maleate is a class II anti-arrhythmic agent that functions as a non-selective β-adrenoceptor antagonist, a vasodilator and an anti-glaucoma agent. Studies have reported that timolol maleate can be used for the treatment of infantile hemangioma (IH)7.

Features and Benefits

This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Preparation Note

Timolol maleate is soluble in ethanol and water.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
M110C6848E6888
form

powder

form

powder

form

powder

form

powder

assay

≥98% (TLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98.0% (HPLC)

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100

solubility

H2O: soluble, ethanol: soluble

solubility

H2O: >3 mg/mL, clear (warmed)

solubility

DMSO: 32 mg/mL, H2O: insoluble

solubility

methanol: ≥50 mg/mL, clear, colorless to yellow

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

originator

-

originator

-

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

color

white to off-white

color

white to dark brown

color

white to beige

color

white to off-white


pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Ajith Chakkittakandiyil et al.
Pediatric dermatology, 29(1), 28-31 (2011-12-14)
Therapeutic options for superficial infantile hemangiomas (IH) are limited. Recently, timolol maleate gel, a topical nonselective beta-blocker, has been reported as a potentially effective treatment for superficial IH. This study is an extension of a previously published pilot study designed
Verena Prokosch et al.
PloS one, 8(2), e49730-e49730 (2013-03-08)
Optic nerve atrophy caused by abnormal intraocular pressure (IOP) remains the most common cause of irreversible loss of vision worldwide. The aim of this study was to determine whether topically applied IOP-lowering eye drugs affect retinal ganglion cells (RGCs) and
G C Chiou et al.
Journal of ocular pharmacology, 8(3), 183-190 (1992-01-01)
An ocular hypertensive rabbit eye model was used to study the effect of L- and D-timolol on retinal and choroidal blood flow. Ocular hypertension was induced artificially by raising the intraocular pressure to 40 mmHg which reduced the ocular blood



Global Trade Item Number

SKUGTIN
T6394-1G04061837371400
T6394-250MG04061832094076
T6394-100MG04061832094069

Questions

Reviews

No rating value

Active Filters