Skip to Content
Merck

Skip To

T5391

apo-Transferrin human

γ-irradiated, powder, BioXtra, suitable for cell culture, ≥98%

Synonym(s):

Human transferrin, Siderophilin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
234-318-8
NACRES:
NA.75
MDL number:
eCl@ss:
34058011
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

sterility

γ-irradiated

product line

BioXtra

assay

≥98%

form

powder

mol wt

76-81 kDa

concentration

~25 mM

technique(s)

cell culture | mammalian: suitable

impurities

HIV and HBsAg, source material tested negative, iron, essentially free

solubility

H2O: 50 mg/mL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TF(7018)

General description

apo-Transferrin (aTF) human can be supplemented with iron or used to bind free iron present in media. It can be used to transport iron into mammalian cells.

Application

apo-Transferrin human has been used as a supplement:
  • in Dulbecco′s modified Eagle medium/HamF12 (DMEM/F-12) to culture human adrenocortical carcinoma cells[1]
  • in DMEM/F-12 to culture neural precursor cells from mice[2]
  • in collecting medium NCSU23 to culture preantral follicles from porcine ovaries[3]

Biochem/physiol Actions

apo-Transferrin (aTF) is an endogenous iron chelator and shows protective effects in the host by scavenging free iron and preventing the formation of toxic reactive oxygen species (ROS). It shows a therapeutic effect against thalassemia.[4] After the release of iron, receptor-bound apo-transferrin reaches the cell surface instead of being transported to lysosomes for degradation. The cell surface has a neutral pH that promotes the release of apo-transferrin from its receptor, where it can again circulate and bind additional iron to undergo further rounds of iron delivery to cells.

Other Notes

Non-heme iron-transport protein.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T4382T2252T8158
form

powder

form

powder

form

powder

form

powder

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

sterility

γ-irradiated

sterility

-

sterility

-

sterility

-

solubility

H2O: 50 mg/mL

solubility

H2O: 50 mg/mL

solubility

H2O: 50 mg/mL

solubility

H2O: 50 mg/mL

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

ambient


Still not finding the right product?

Explore all of our products under apo-Transferrin human


Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Ji Wu et al.
Journal of assisted reproduction and genetics, 24(9), 419-424 (2007-07-20)
There has been controversy over the role of FSH in the regulation of preantral follicle development. LH is a survival and differentiation factor that increases oocyte maturation in FSH-supplemented cultures of mouse preantral follicles. However, little information exists on the
Hiroshi Kawabata
Free radical biology & medicine, 133, 46-54 (2018-07-04)
In vertebrates, transferrin (Tf) safely delivers iron through circulation to cells. Tf-bound iron is incorporated through Tf receptor (TfR) 1-mediated endocytosis. TfR1 can mediate cellular uptake of both Tf and H-ferritin, an iron storage protein. New World arenaviruses, which cause
Miwako Yamamiya et al.
Biochemical and biophysical research communications, 513(4), 841-845 (2019-04-21)
Microglia are central nervous system-resident immune cells that play a crucial role in brain development by interacting with neural precursor cells (NPCs). It has been reported that microglia regulate the number of NPC by phagocytosis, inducing apoptosis, and promoting proliferation.



Global Trade Item Number

SKUGTIN
T5391-10MG04061837347474

Questions

1–2 of 2 Questions  
  1. What is the carbohydrate content of human transferrin?

    1 answer
    1. We have found the information on transferrin in a reference book:The Plasma Proteins, F.W. Putnam, ed, volume 2, chapter 4, Table II.According to the table, Transferrin is 6 percent carbohydrate, citing G.A. Jamieson, J. Biol. Chem. 240(7), 2914-2920 (1965).The table also gives information on the percentage of each type of monosaccharide that is present.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters