Skip to Content
Merck

Skip To

T5192

Anti-γ-Tubulin Antibody

rabbit polyclonal

Synonym(s):

Anti-γ-Tubulin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, WB
Citations:
240
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-γ-Tubulin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution

biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 48 kDa

species reactivity

human, chicken

packaging

antibody small pack of 25 μL

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

indirect immunofluorescence: 1:1,000 using methanol/acetone-fixed chicken fibroblasts cells, western blot: 1:1,000 using whole cell extract human epidermal carcinoma A431 cell line

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

γ-Tubulin, mapped on human chromosome 17q21.2, codes for a member of the tubulin family. It is localized to the microtubule organizing centres. γ-Tubulin consists of two isoforms tubulin γ 1 (TUBG1) and tubulinγ 2 (TUBG2) with 97.3% amino acid identity. In addition to these two isoforms, γ-tubulin pseudogene (TUBG1P) is also been identified on human chromosome 17. Human TUBG1 and TUBG2 transcripts are widely expressed in preimplantation embryos and the brain, respectively. γ-tubulin is a component of γ-tubulin ring complex (γ-TuRC), which has roughly the same diameter as a microtubule. γ-Tubulin does not polymerize with α-tubulin/β-tubulin, but instead it is localized to the centrosome and to the cytoplasm.

Immunogen

synthetic peptide corresponding to the N-terminal region of human γ-tubulin (amino acids 38-53, with C-terminally added lysine) conjugated to KLH. This sequence is specific for γ-tubulin and not found in other members of the tubulin family such as α− β−, δ and ε− tubulins. This sequence is identical in mouse and rat γ-tubulin and highly conserved among species (Drosophila, Aspergillus, and yeast γ-tubulin).

Application

Anti-γ-Tubulin antibody produced in rabbit has been used in following studies:
  • Immunocytochemistry.
  • Immunofluorescence.
  • Immunoblotting.
  • Staining in microscopy.
  • Chromatin immunoprecipitation (ChIP).

Biochem/physiol Actions

γ-Tubulin nucleates microtubule assembly throughout the mammalian cell cycle in vivo. In Aspergillus nidulans, γ-tubulin facilitates attachment of microtubules to the spindle pole body, nuclear division and microtubule assembly. Ubiquitination of γ-tubulin by breast cancer 1 protein (BRCA1) is a crucial step in the regulation of centrosome number. Overexpression of γ-tubulin is observed in lung cancer. Tubulin γ 2 (TUBG2) plays a vital role in cell growth. Aberration in the γ-tubulin gene alters microtubule assembly. The expression levels of γ-tubulin can be considered as an important prognostic indicator for patients with astrocytomas.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T3559T3320T5293
antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

antibody form

IgG fraction of antiserum

antibody form

ascites fluid

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

mouse

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

form

buffered aqueous solution

form

buffered aqueous solution

form

buffered aqueous solution

form

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters