Skip to Content
Merck

Skip To

T4264

Tetrahydrozoline hydrochloride

≥98% (HPLC)

Synonym(s):

4,5-Dihydro-2-(1,2,3,4-tetrahydro-1-naphthalenyl)-1H-imidazole monohydrochloride, Tetryzoline hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H16N2 · HCl
CAS Number:
Molecular Weight:
236.74
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
208-329-3
MDL number:
Assay:
≥98% (HPLC)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

solubility

H2O: freely soluble, alcohol: freely soluble, chloroform: very slightly soluble, diethyl ether: insoluble

originator

Novartis

SMILES string

Cl.C1CC(C2=NCCN2)c3ccccc3C1

InChI

1S/C13H16N2.ClH/c1-2-6-11-10(4-1)5-3-7-12(11)13-14-8-9-15-13;/h1-2,4,6,12H,3,5,7-9H2,(H,14,15);1H

InChI key

BJORNXNYWNIWEY-UHFFFAOYSA-N

Biochem/physiol Actions

Tetrahydrozoline is an α-adrenergic agonist and an imidazoline derivative.[1][2] It plays a crucial role in the constriction of conjunctival blood vessels.[2] Tetrahydrozoline is a common component of nasal and eye drop formulations.[2]

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1652001Y0000460T6950
assay

≥98% (HPLC)

assay

-

assay

-

assay

≥98% (HPLC)

Quality Level

200

Quality Level

-

Quality Level

-

Quality Level

100

solubility

H2O: freely soluble, chloroform: very slightly soluble, alcohol: freely soluble, diethyl ether: insoluble

solubility

-

solubility

-

solubility

H2O: >20 mg/mL

originator

Novartis

originator

-

originator

-

originator

Acorda Therapeutics

Gene Information

human ... ADRA1A(148), ADRA1B(147), ADRA1D(146), ADRA2A(150), ADRA2B(151), ADRA2C(152)

Gene Information

human ... ADRA1A(148), ADRA1B(147), ADRA1D(146), ADRA2A(150), ADRA2B(151), ADRA2C(152)

Gene Information

human ... ADRA1A(148), ADRA1B(147), ADRA1D(146), ADRA2A(150), ADRA2B(151), ADRA2C(152)

Gene Information

human ... ADRA2A(150), ADRA2B(151), ADRA2C(152)


Still not finding the right product?

Explore all of our products under Tetrahydrozoline hydrochloride


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters