Skip to Content
Merck

Skip To

SML2372

β-Muricholic Acid

≥98% (HPLC), farnesoid X receptor antagonist, powder

Synonym(s):

β-MCA, (3a,5b,6b,7b)-3,6,7-trihydroxy-cholan-24-oic acid, 5β-Cholanic acid-3α,6β,7β-triol, beta-Muricholic Acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C24H40O5
CAS Number:
Molecular Weight:
408.57
UNSPSC Code:
12352106
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

β-Muricholic Acid, ≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +59 to +65°, c = 0.5 in methanol

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O[C@H]1[C@H]2[C@@](C3C([C@H]1O)C4[C@@](C(CC4)C(CCC(=O)O)C)(CC3)C)(CC[C@H](C2)O)C

InChI

1S/C24H40O5/c1-13(4-7-19(26)27)15-5-6-16-20-17(9-11-23(15,16)2)24(3)10-8-14(25)12-18(24)21(28)22(20)29/h13-18,20-22,25,28-29H,4-12H2,1-3H3,(H,26,27)/t13?,14-,15?,16?,17?,18+,20?,21+,22-,23-,24-/m1/s1

InChI key

DKPMWHFRUGMUKF-CWNVCPLQSA-N

Application

β-Muricholic Acid has been used as a bile acid to study its effects on the expression of pluripotency, stemness, and inflammatory genes in primary mouse hepatocytes (PMH) and tumor-initiating stem-like cells(TICs).[1] It has also been used as a bile acid to study its effects on gut transit ingnotobiotic or conventional mouse models.[2]

Biochem/physiol Actions

β-Muricholic Acid is a naturally occurring trihydroxy hydrophilic bile acid that is produced in the liver of murine. It serves as a biliary cholesterol-desaturating agent and aids in the dissolution of cholesterol gallstones.[3]
β-Muricholic acid (β-MCA) is a natural murine-specific bile acid. β-Muricholic acid has been shown to reduce the absorption of cholesterol. It also has some activity as a farnesoid X receptor antagonist.
Murine-specific bile acid, farnesoid X receptor antagonist

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2352700233P700232P
form

powder

form

powder

form

powder

form

powder

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

>99% (TLC)

assay

>99% (TLC)

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

−20°C

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

-

solubility

-

color

white to beige

color

white to beige

color

-

color

-

optical activity

[α]/D +59 to +65°, c = 0.5 in methanol

optical activity

-

optical activity

-

optical activity

-


Still not finding the right product?

Explore all of our products under β-Muricholic Acid


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Laura Conde de la Rosa et al.
Journal of hepatology, 74(6), 1429-1441 (2021-01-31)
Besides their physiological role in bile formation and fat digestion, bile acids (BAs) synthesised from cholesterol in hepatocytes act as signalling molecules that modulate hepatocellular carcinoma (HCC). Trafficking of cholesterol to mitochondria through steroidogenic acute regulatory protein 1 (STARD1) is
Kyosuke Fujita et al.
Lipids, 52(4), 335-344 (2017-03-21)
The farnesoid X receptor (FXR) is a major nuclear receptor of bile acids; its activation suppresses sterol regulatory element-binding protein 1c (SREBP1c)-mediated lipogenesis and decreases the lipid contents in the liver. There are many reports showing that the administration of
Ylva Bonde et al.
PloS one, 11(1), e0147772-e0147772 (2016-01-30)
High endogenous production of, or treatment with muricholic bile acids, strongly reduces the absorption of cholesterol. Mice abundant in muricholic bile acids may therefore display an increased resistance against dietary induced weight gain, steatosis, and glucose intolerance due to an



Global Trade Item Number

SKUGTIN
SML2372-5MG04061838811646
SML2372-25MG04061838811639

Questions

Reviews

No rating value

Active Filters