Skip to Content
Merck

Skip To

SML1289

Desferrithiocin sodium salt

≥98% (HPLC)

Synonym(s):

(4S)-4,5-Dihydro-2-(3-hydroxy-2-pyridinyl)-4-methyl-4-Thiazolecarboxylic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C10H9N2O3S · xNa+
CAS Number:
Molecular Weight:
237.26 (free acid basis)
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

faintly yellow to dark yellow

solubility

H2O: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

C[C@]1(C([O-])=O)CSC(C2=C(C=CC=N2)O)=N1

InChI

1S/C10H10N2O3S/c1-10(9(14)15)5-16-8(12-10)7-6(13)3-2-4-11-7/h2-4,13H,5H2,1H3,(H,14,15)/p-1/t10-/m1/s1

InChI key

MJWAGSZZOQMRNY-SNVBAGLBSA-M

Biochem/physiol Actions

Desferrithiocin is an iron chelator.
Iron chelator.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML0289P0039SML0660
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

solid

form

powder

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

solubility

H2O: 15 mg/mL, clear

solubility

H2O: ≥5 mg/mL

solubility

DMSO: 10 mg/mL, clear (warmed)

solubility

DMSO: 15 mg/mL, clear

color

faintly yellow to dark yellow

color

faintly yellow to dark yellow

color

red

color

white to beige


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters