Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Empirical Formula (Hill Notation):
C27H38N6O9
CAS Number:
Molecular Weight:
590.63
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
MDL number:
Pricing and availability is not currently available.
Product Name
N-Succinyl-Ala-Ala-Pro-Leu p-nitroanilide, protease substrate
Quality Segment
form
solid
storage temp.
−20°C
SMILES string
CC(C)CC(NC(=O)C1CCCN1C(=O)C(C)NC(=O)C(C)NC(=O)CCC(O)=O)C(=O)Nc2ccc(cc2)N(=O)=O
InChI
1S/C27H38N6O9/c1-15(2)14-20(25(38)30-18-7-9-19(10-8-18)33(41)42)31-26(39)21-6-5-13-32(21)27(40)17(4)29-24(37)16(3)28-22(34)11-12-23(35)36/h7-10,15-17,20-21H,5-6,11-14H2,1-4H3,(H,28,34)(H,29,37)(H,30,38)(H,31,39)(H,35,36)
InChI key
PTHRPHGMGFMCSS-UHFFFAOYSA-N
General description
Substrate for elastase
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 200 | Quality Level - | Quality Level 200 | Quality Level 300 |
| form solid | form solid | form solid | form powder |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



