Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Linear Formula:
CH3(CH2)16COOC2H5
CAS Number:
Molecular Weight:
312.53
UNSPSC Code:
12352211
NACRES:
NA.25
PubChem Substance ID:
EC Number:
203-887-4
Beilstein/REAXYS Number:
1788183
MDL number:
Pricing and availability is not currently available.
biological source
plant
Quality Level
assay
≥99% (capillary GC)
form
powder
bp
213-215 °C/15 mmHg (lit.)
mp
34-38 °C (lit.)
functional group
ester
lipid type
saturated FAs
shipped in
wet ice
storage temp.
2-8°C
SMILES string
CCCCCCCCCCCCCCCCCC(=O)OCC
InChI
1S/C20H40O2/c1-3-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20(21)22-4-2/h3-19H2,1-2H3
InChI key
MVLVMROFTAUDAG-UHFFFAOYSA-N
Disclaimer
Low melting solid
1 of 1
This Item | |||
|---|---|---|---|
| functional group ester | functional group ester | functional group ester | functional group ester |
| assay ≥99% (capillary GC) | assay ≥98.0% (GC) | assay ~99% (GC) | assay ≥99% (capillary GC) |
| biological source plant | biological source synthetic | biological source - | biological source - |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. −20°C | storage temp. 2-8°C |
| shipped in wet ice | shipped in ambient | shipped in ambient | shipped in ambient |
| form powder | form powder | form powder | form powder |
Still not finding the right product?
Explore all of our products under Ethyl stearate
Storage Class
11 - Combustible Solids
wgk
WGK 2
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S Aliaskarisohi et al.
The journal of physical chemistry. B, 115(40), 11631-11637 (2011-08-31)
In a previous work, Muruganathan and Fischer observed laser-induced local collapse of a methyl stearate monolayer. These experiments opened the possibility of studying the collapse mechanism in a highly controlled manner, since the laser intensity can be easily varied and
The dual role of macrophages in the sporozoite-induced malaria infection. A hypothesis.
J P Verhave et al.
International journal of nuclear medicine and biology, 7(2), 149-156 (1980-01-01)
Yan-Bing Zhang et al.
Journal of Asian natural products research, 8(1-2), 119-123 (2006-06-07)
A new phenolic glycoside (1), mahkoside A, together with six known compounds including mangiferin (2), kaempferol-3-O-beta-D-glucoside (3), dodecanoic acid (4), palmitic acid (5) ethyl stearate (6) and sucrose (7), were isolated from the pit of Mahkota dewa. Their structures were
Global Trade Item Number
| SKU | GTIN |
|---|---|
| S8269-5G | 04061836960681 |
| S8269-25G | 04061836960674 |



