Skip to Content
Merck

Skip To

S7900

Saponin from quillaja bark

Sapogenin content ≥10 %

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12161902
EC Number:
232-462-6
NACRES:
NA.32
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

non-ionic

Quality Level

composition

Sapogenin content, ≥10%

technique(s)

HPLC: suitable, protein quantification: suitable

sulfated ash

<20%

SMILES string

O1[C@H]([C@@H]([C@H]([C@@H]([C@H]1CO)O)O)O)O[C@H]2[C@@H](O[C@@H]([C@H]([C@@H]2O)O[C@@H]8O[C@@H]([C@H]([C@@H]([C@H]8O)O)O)CO)C(=O)O)O[C@@H]3[C@@](C4[C@@](C5[C@]([C@@]6(C[C@H]([C@]7([C@H]([C@@H](C(CC7C6=CC5)(C)C)OC(=O)\C(=C/C)\C)OC(=O)C)CO)O)C)(CC4)C)(CC3)C

InChI key

AXNVHPCVMSNXNP-ZELRDNAQSA-N

General description

Saponins are natural steroidal glycoside (terpenes).

Application

Saponin from Quillaja bark has been used:
  • to stain CD4+ and CD8+ T-cells
  • to lyse trophozoite-infected red blood cells
  • in immunohistochemical staining

Biochem/physiol Actions

Saponins are associated with many biological and pharmacological activities. It is known to stimulate immune response in mammals. Hence it is used as vaccine adjuvants. Saponins possesses antiviral, antifungal, antitumor, anti-inflammatory, hypoglycemic and hypocholesterolemic effects.
Potent hemolytic when injected i.v.; surfactant that enhances penetration of proteins and other macromolecules through cell membranes; it also has been used as an adjuvant for vaccines.
Saponins solubilize the low density lipoprotein (LDL) fraction of cholesterol in micelles, thus lowering their concentration. Saponin from Quillaja bark is used as a flavoring agent in food and beverages. This saponin has been shown to enhance immune-cell proliferation in vitro.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S452184510SAE0073
technique(s)

HPLC: suitable, protein quantification: suitable

technique(s)

HPLC: suitable, protein quantification: suitable

technique(s)

HPLC: suitable, protein quantification: suitable

technique(s)

HPLC: suitable, LC/MS: suitable, protein quantification: suitable

description

non-ionic

description

non-ionic

description

-

description

non-ionic

Quality Level

200

Quality Level

300

Quality Level

300

Quality Level

200

sulfated ash

<20%

sulfated ash

≤10%

sulfated ash

-

sulfated ash

≤10%

composition

Sapogenin content, ≥10%

composition

Sapogenin content, 20-35%

composition

Sapogenin, 8-25%

composition

Sapogenin content, 20-35%


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Samantha Reiss et al.
PloS one, 12(10), e0186998-e0186998 (2017-10-25)
The identification and study of antigen-specific CD4 T cells, both in peripheral blood and in tissues, is key for a broad range of immunological research, including vaccine responses and infectious diseases. Detection of these cells is hampered by both their
Metabolite profiling of the intraerythrocytic malaria parasite Plasmodium falciparum by (1)H NMR spectroscopy.
Teng R
Nmr in Biomedicine, 22(3), 292-302 (2009)
Effect of testosterone on insulin stimulated IRS1 Ser phosphorylation in primary rat myotubes--a potential model for PCOS-related insulin resistance.
Allemand MC
PLoS ONE, 4(1), e4274-e4274 (2009)



Global Trade Item Number

SKUGTIN
S7900-100G04061835559138
S7900-25G04061835559145

Questions

1–3 of 3 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. Is this standard suitable for testing saponin content in Quillaja saponaria extracts?

    1 answer
    1. S7900 is Reagent grade of Saponin from quillaja bark. As per our policy: https://www.sigmaaldrich.com/technical-documents/technical-article/analytical-chemistry/calibration-qualification-and-validation/how-to-choose-the-correct-reference-material-quality-grade
      This product is not characterized for use as reference materials. As shown in Table 4, at least Reference Material grade is qualified for Content or Assay use. Reagent-grade products may only be suitable for identity tests.

      Helpful?

Reviews

No rating value

Active Filters