Skip to Content
Merck

Skip To

S4526

Salinomycin

from Streptomyces albus, ≥98% (HPLC), powder, MDRP-1 inhibitor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C42H70O11
CAS Number:
Molecular Weight:
751.00
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
EC Number:
258-290-1
Assay:
≥98% (HPLC)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Salinomycin, from Streptomyces albus, ≥98% (HPLC)

biological source

Streptomyces albus

Quality Segment

assay

≥98% (HPLC)

mp

112.5-113.5 °C (lit.)

antibiotic activity spectrum

neoplastics

mode of action

cell membrane | interferes

storage temp.

2-8°C

SMILES string

CC[C@H]([C@H]1CC[C@H](C)[C@@H](O1)[C@@H](C)[C@H](O)[C@H](C)C(=O)[C@H](CC)[C@H]2O[C@@]3(O[C@@]4(CC[C@](C)(O4)[C@H]5CC[C@](O)(CC)[C@H](C)O5)[C@H](O)C=C3)[C@H](C)C[C@@H]2C)C(O)=O

InChI

1S/C42H70O11/c1-11-29(38(46)47)31-15-14-23(4)36(50-31)27(8)34(44)26(7)35(45)30(12-2)37-24(5)22-25(6)41(51-37)19-16-32(43)42(53-41)21-20-39(10,52-42)33-17-18-40(48,13-3)28(9)49-33/h16,19,23-34,36-37,43-44,48H,11-15,17-18,20-22H2,1-10H3,(H,46,47)/t23-,24-,25+,26-,27-,28-,29+,30-,31+,32+,33+,34+,36+,37-,39-,40+,41-,42-/m0/s1

InChI key

KQXDHUJYNAXLNZ-XQSDOZFQSA-N

General description

Chemical structure: polyether

Application

Salinomycin was used as a standard in the development of LC-MS/MS method for detection of residual coccidiostats in egg and muscle samples.[1] It was used to screen out CD44+CD24- mesenchymal-like subpopulation within breast carcinomas.[2]

Biochem/physiol Actions

Salinomycin produced by Streptomyces albus is a carboxylic polyether ionophore with antibiotic and anti-cancer properties. It induces cell death in some types of cancer cells such as breast, lung, gastric cancer, leukemia and osteosarcoma. Salinomycin inhibits multidrug resistance protein 1 and induces apoptosis by the generation of reactive oxygen species that cause DNA damage and inactivation of Stat3.[3][4] Use of salinomycin as antibiotic results in lowered spermatozoa count and motility and decreased steroidogenesis in mice.[5]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
V363994675V0627
assay

≥98% (HPLC)

assay

≥90% (HPLC)

assay

≥99.0% (TLC)

assay

≥90% (HPLC), ≥98% (TLC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

300

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

biological source

Streptomyces albus

biological source

Streptomyces sp.

biological source

-

biological source

-

antibiotic activity spectrum

neoplastics

antibiotic activity spectrum

Gram-positive bacteria, parasites, viruses

antibiotic activity spectrum

mycobacteria

antibiotic activity spectrum

Gram-positive bacteria, parasites, viruses

mp

112.5-113.5 °C (lit.)

mp

-

mp

187-190 °C

mp

-


Still not finding the right product?

Explore all of our products under Salinomycin


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters