Skip to Content
Merck

Skip To

P9416

TWEEN® 20

Molecular Biology, viscous liquid

Synonym(s):

Polyethylene glycol sorbitan monolaurate, Polyoxyethylenesorbitan monolaurate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12161902
NACRES:
NA.31
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

corn, plant (Palm), plant (coconut), wheat

grade

Molecular Biology

description

non-ionic

assay

>40% (GC)

form

viscous liquid

mol wt

~1228

composition

lauric acid, ≥40% (balance primarily myristic, palmitic, and stearic acids)

concentration

≥40.0% (GC)

impurities

RNAse, none detected, endonuclease, none detected, exonuclease, none detected

refractive index

n20/D 1.468 (lit.)

CMC

60 mg/L

solubility

water: 100 mg/mL, clear, colorless to yellow

density

1.095  g/mL at 25 °C ((lit.)), 1.095 g/mL at 25 °C (lit.)

suitability

suitable for molecular biology

foreign activity

Endonuclease-exonuclease and RNase, none detected

SMILES string

CCCCCCCCCCCC(=O)OCCOCC(C1C(C(CO1)OCCO)OCCO)OCCO

InChI

1S/C26H50O10/c1-2-3-4-5-6-7-8-9-10-11-24(30)34-19-18-31-20-22(32-15-12-27)26-25(35-17-14-29)23(21-36-26)33-16-13-28/h22-23,25-29H,2-21H2,1H3

InChI key

HMFKFHLTUCJZJO-UHFFFAOYSA-N

General description

Tween® 20 is a nonionic surfactant and detergent widely applied in molecular biology, biochemical, and life science research. Its predominant use involves the solubilization of membrane proteins during the isolation of membrane-protein complexes. Additionally, it serves as a component in various washing, blocking, dilution, and stabilization buffers. Additionally, it functions as a reagent for immunohistochemistry, encompassing techniques like Western blotting and ELISAs. In these applications, Tween 20 plays crucial roles such as preventing non-specific antibody binding, minimizing background staining, and ensuring the even distribution of reagents.

Surfactants, possessing both hydrophilic and hydrophobic components, fall into two main categories: ionic, which ionize in water (anionic, cationic, or ampholytic), and nonionic, which do not ionize. The discussed product is a nonionic surfactant with diverse applications such as emulsification, dispersion, solubilization, and detergent use. Additionally, it can serve as a solubilizer for membrane proteins.
Tween®20 is a polyoxyethylene sorbitol ester that belongs to the polysorbate family. It is a nonionic detergent having a molecular weight of 1,225 daltons, assuming 20 ethylene oxide units, 1 sorbitol, and 1 lauric acid as the primary fatty acid. The ethylene oxide subunits are responsible for the hydrophilic nature of the surfactant, while the hydrocarbon chains provide the hydrophobic environment. Sorbitol forms the backbone ring to which the ethylene oxide polymers are attached.

Application

Non-ionic detergent
TWEEN 20 is widely used in biochemical applications. It has been used:
  • As an emulsifying agent for the preparation of stable oil-in-water emulsions
  • In pre-extraction of membranes to remove peripheral proteins (used at 2% for extraction of membrane-bound proteins)
  • As a blocking agent for membrane based immunoassays at a typical concentration of 0.05%
  • For lysing mammalian cells at a concentration of 0.005 to 0.5%
  • Along with PBS in the dilution of antibodies in immunohistochemistry technique
  • For washing cells in FISH (Fluorescence in situ hybridization) technique[1]

Features and Benefits

  • Highly versatile surfactant for your Life Science and Biochemical research
  • Suitable for sensitive molecular biology research applications
  • Tested for Endonuclease, exonuclease and Rnase′s
  • Excellent solubilising, dispersing and emulsifying agent

Preparation Note

TWEEN 20 is miscible in water (100mg/mL), yielding a clear, yellow solution. It is also miscible with alcohol, dioxane, and ethyl acetate. TWEEN 20 is insoluble in liquid paraffin and fixed oils.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Disclaimer

TWEEN 20 may not be stable to autoclaving, particularly with metal cations in buffer solutions. Autoclaving is not recommended without testing for changes in properties. TWEEN 20 is heat sensitive and will darken when exposed to elevated temperatures. Polysorbates have been reported to be incompatible with alkalis, heavy metal salts, phenols, and tannic acid. They may also reduce the activity of many preservatives. Aqueous solutions of polysorbates undergo autoxidation during storage, with changes being catalyzed by light, increased temperature, and copper sulfate.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P1379P228793773
grade

Molecular Biology

grade

-

grade

-

grade

-

suitability

suitable for molecular biology

suitability

-

suitability

-

suitability

-

assay

>40% (GC)

assay

>40% (GC)

assay

-

assay

-

form

viscous liquid

form

viscous liquid

form

viscous liquid

form

viscous liquid

density

1.095  g/mL at 25 °C ((lit.)), 1.095 g/mL at 25 °C (lit.)

density

1.095 g/mL at 25 °C (lit.)

density

1.095 g/mL at 25 °C (lit.), 1.1 g/cm3 at 25 °C (lit.)

density

1.11 g/mL at 20 °C, 1.095 g/mL at 25 °C (lit.)

biological source

corn, plant (coconut), plant (Palm), wheat

biological source

-

biological source

-

biological source

-


Still not finding the right product?

Explore all of our products under TWEEN® 20


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

527.0 °F - Pensky-Martens closed cup

flash_point_c

275 °C - Pensky-Martens closed cup

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Reynolds, J. E. F., (eds.)
Martindale: The Extra Pharmacopoeia, 1030-1030 (1993)
Telomere analysis by fluorescence in situ hybridization and flow cytometry.
Magnus H, et al.
Nucleic Acids Research, 26(16), 3651-3656 (1998)
The role of transforming growth factor beta isoforms in tendon-to-bone healing
Kim H M, et al.
Connective tissue research, 52(2), 87-98 (2011)



Global Trade Item Number

SKUGTIN
P9416-50ML04061838258786
P9416-100ML04061838258779

Questions

1–7 of 7 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  3. How should diluted Tween20 (1% Tween20 in water) be stored and for how long? Should it be stored at 4°C?

    1 answer
    1. There are no official recommendations available for storing diluted Tween20. ​However, some literature advises to store the dilute solution at 4°C, ensuring it is protected from light. If the solution becomes cloudy, it is recommended to discard it. Additionally, it is advisable to use dilute solutions within one week.

      Helpful?

  4. is this tween 20 is sterile

    1 answer
    1. This product is not sterile. Currently, there are no sterile Tween 20 products available. Please see the link below to review the product datasheet for additional information:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/128/958/p9416pis.pdf

      Helpful?

  5. добры дзень. Буду ўдзячны, калі вы паведаміце мне канцэнтрацыі TWEEN20 (P9416) і TWEEN80 (P1754). Таксама дайце мне ведаць, W/V% або V/V%D. Дзякуй

    1 answer
    1. Both products are essentially liquid detergents. These are neat liquids and the concentration is considered 100 %.

      Helpful?

  6. Is this in a light-sensitive bottle?

    1 answer
    1. This product is packaged in a white HDPE bottle, which is considered light-protective.
      See the image below:

      Helpful?

  7. 안녕하세요. Tween20 (P9416)과 Tween80 (P1754)의 농도를 알려주시면 감사하겠습니다. w/v%인지 v/v%d인지도 안내 부탁드립니다. 감사합니다.

    1 answer
    1. Both products are essentially liquid detergents. These are neat liquids and the concentration is considered 100%.

      Helpful?

Reviews

No rating value

Active Filters