Skip to Content
Merck

Skip To

P8044

Proteinase K from Tritirachium album

≥3.0 unit/mg solid, lyophilized powder

Synonym(s):

Endopeptidase K

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
EC Number:
254-457-8
NACRES:
NA.54
EC Number:
Specific activity:
≥3.0 unit/mg solid
Biological source:
maize, microbial (T.album
T. ALBUM), plant seeds (barley), soybean, yeast
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

maize, microbial (T.album
T. ALBUM), plant seeds (barley), soybean, yeast

form

lyophilized powder

specific activity

≥3.0 unit/mg solid

mol wt

28.93 kDa

composition

Protein, ≥15% biuret

technique(s)

DNA extraction: suitable

application(s)

diagnostic assay manufacturing

storage temp.

−20°C

Application

Proteinase K is useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA. It is used for the removal of endotoxins bound to cationic proteins such as lysozyme and ribonuclease A. It is also useful for the isolation of hepatic, yeast, and mung bean mitochondria and is used to determine enzyme localization on membranes. Furthermore, it is used for the treatment of paraffin embedded tissue sections to expose antigen binding sites and for digestion of proteins from brain tissue samples. Product P8044 is provided as a lyophilized powder[1].
Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA.
Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA.
Removes endotoxins that bind to cationic proteins such as lysozyme and ribonuclease A.
Reported useful for the isolation of hepatic, yeast, and mung bean mitochondria
Determination of enzyme localization on membranes
Treatment of paraffin embedded tissue sections to expose antigen binding sites for antibody labeling.
Digestion of proteins from brain tissue samples for prions in Transmissible Spongiform Encephalopathies (TSE) research.
The enzyme from Sigma has been used to degrade complex I (NADH:ubiquinone oxidoreductase) prior to extraction of ubiquinone from Yarrowia lipolytica.[2] It has been used to examine the effect of Proteinase k on Pythium ultimum.[3] It has also been used to maximize the variety of peptide bonds hydrolyzed in the sediment slurry without autolytic production of amino acids from the enzyme itself.

Biochem/physiol Actions

Proteinase K is a stable and highly reactive serine protease. Evidence from crystal and molecular structure studies indicates the enzyme belongs to the subtilisin family with an active-site catalytic triad (Asp39-His69-Ser224). It is stable in a broad range of environments: pH, buffer salts, detergents (SDS), and temperature. In the presence of 0.1-0.5% SDS, proteinase K retains activity and will digest a variety of proteins and nucleases in DNA preparations without compromising the integrity of the isolated DNA.
Proteinase K is highly active towards native proteins. It has a broad specificity and degrades many proteins even in the native state. It mainly cleaves the peptide bond adjacent to the carboxyl group of aliphatic and aromatic amino acids with blocked α-amino groups.[4][5] The optimum pH is between 7.5-9.0[4] and the isoelectric point is 8.9.[4] Ca2+ (1-5 mM) is required for activation. Proteinase K is inhibited by diisopropyl fluorophosphate (DFIP), and phenylmethanesulfonyl fluoride (PMSF).

Other Notes

One unit will hydrolyze urea-denatured hemoglobin to produce color equivalent to 1.0 μmole of tyrosine per min at pH 7.5 at 37 °C (color by Folin-Ciocalteu reagent).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SRE0047SRE0005P6556
specific activity

≥3.0 unit/mg solid

specific activity

≥30 units/mg protein

specific activity

-

specific activity

≥30 units/mg protein

biological source

maize, plant seeds (barley), yeast, microbial (T.album
T. ALBUM), soybean

biological source

fungus (Tritirachium album)

biological source

microbial

biological source

-

technique(s)

DNA extraction: suitable

technique(s)

DNA extraction: suitable, RNA extraction: suitable

technique(s)

DNA extraction: suitable, RNA extraction: suitable

technique(s)

DNA extraction: suitable

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

diagnostic assay manufacturing

application(s)

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

mol wt

28.93 kDa

mol wt

28,930 Da, 28.93 kDa

mol wt

28.93 kDa

mol wt

28.93 kDa


Still not finding the right product?


pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters