Skip to Content
Merck

Skip To

P7405

Poly-ᴅ-Lysine Hydrobromide

synthetic, mol wt >300,000, powder, γ-irradiated, suitable for cell culture, BioXtra

Synonym(s):

PDL

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
D-Lys-(D-Lys)n-D-Lys · xHBr
CAS Number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.75
MDL number:
Biological source:
synthetic (chemical)
Form:
lyophilized powder
Technique(s):
cell culture | mammalian: suitable
Concentration:
0.016—0.032 mmol lysine
Mol wt:
>300,000
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Poly-D-lysine hydrobromide, mol wt >300,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture

biological source

synthetic (chemical)

Quality Level

sterility

γ-irradiated

product line

BioReagent, BioXtra

form

lyophilized powder

mol wt

>300,000

packaging

pkg of 5 mg

concentration

0.016—0.032 mmol lysine

technique(s)

cell culture | mammalian: suitable

surface coverage

4 μg/cm2

solubility

H2O: soluble 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(C)[C@@](NC)([H])CCCCN.[Br]

InChI

1S/C6H14N2O2.BrH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H

InChI key

MEXAGTSTSPYCEP-UHFFFAOYSA-N

General description

Poly-D-Lysine Hydrobromide (PDL) is a cationic polymer composed of lysine residues with an associated hydrobromide molecule per lysine unit. The presence of the hydrobromide molecule introduces a distinctive property to the poly-D-lysine structure allowing the poly-D-lysine to adopt a crystalline form, rendering it soluble in aqueous solutions.

Application

Poly-D-lysine hydrobromide has been used :
  • in preparing the surface of coverslips for cell attachment
  • in coating glass-bottom dishes for live cell imaging of primary cortical neuronal cells
  • in coating 24-well plates for the isolation of primary astrocytes to study the impact of melanoma-derived exosomes on the reprogramming of stromal cells within the metastatic microenvironment
  • for coating 16-well chamber slides to study the effects of maternal immune activation on behavioral impairments and alterations in cytokine and synaptic protein expression in the cerebellum
Poly-D-lysine polymers can be used in preparing surfaces for cell attachment. The D-lysine polymers can also be used with cells that digest poly-L-lysine polymers and cause an excessive uptake of L-lysine.

This product is recommended as a cell culture substratum when using 0.5 - 1.0 mL of a 0.1 mg/mL solution to coat 25 cm2. Lower molecular weight versions of the product are less viscous, but high more molecular weight versions provide more attachment sites per molecule.

Biochem/physiol Actions

Poly-D-Lysine Hydrobromide (PDL) is a nonspecific attachment factor for cells useful in promoting cell adhesion to solid substrates by enhancing electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. PDL increases the number of positively charged cell binding sites post-absorption to the culture surface. PDL coating promotes the attachment of cells to solid surfaces that usually grow in suspension.
Poly-D-lysine (PDL) hydrobromide is a nonspecific attachment factor for cells useful in promoting cell adhesion to solid substrates by enhancing electrostatic interaction between negatively charged ions of the cell membrane and the culture surface. After absorption to the culture surface, poly-D-lysine increases the number of positively charged cell binding sites.

Analysis Note

This product has a molecular weight >300,000 and is cell culture tested and gamma-irradiated. To remove the HBr, dissolve this product in a neutral buffer and dialyze to remove the salts. In general, to use this product as an attachment factor, add 50 mL of sterile tissue culture grade water to 5 mg of poly-lysine, and aseptically coat the surface with 1 mL per 25 cm2 of solution. After 5 minutes, remove the solution through aspiration and thoroughly rinse the surface. Let dry for two hours before introducing cells and medium.

Other Notes

Poly-D-lysine is a positively charged amino acid polymer with approximately one HBr per lysine residue. The hydrobromide allows the poly-D-lysine to be in a crystalline form soluble in water. A small amount of product may be found in the β structure because the HBr interferes with hydrogen bonding between amino and either the carboxyl groups or N or O containing moieties.

Disclaimer

Sterile solutions are stable for up to 2 years when stored at 2-8°C. It should be stored desiccated at -20°C.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P7280P6407P0899
biological source

synthetic (chemical)

biological source

synthetic (organic)

biological source

synthetic (organic)

biological source

-

sterility

γ-irradiated

sterility

γ-irradiated

sterility

γ-irradiated

sterility

-

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

concentration

0.016—0.032 mmol lysine

concentration

0.016—0.032 mmol lysine

concentration

0.016—0.032 mmol lysine

concentration

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

mol wt

>300,000

mol wt

average mol wt 30,000-70,000

mol wt

70,000-150,000

mol wt

70,000-150,000


Still not finding the right product?

Explore all of our products under Poly-D-lysine hydrobromide


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Deficiency of ROS-Activated TRPM2 Channel Protects Neurons from Cerebral Ischemia-Reperfusion Injury through Upregulating Autophagy
Hu X, et al.
Oxidative Medicine and Cellular Longevity (2021)
Maternal Immune Activation Causes Behavioral Impairments and Altered Cerebellar Cytokine and Synaptic Protein Expression
Pendyala G, et al.
Neuropsychopharmacology, 42, 1435?1446-1435?1446 (2017)
Adolfo Alsina et al.
Biochemical and biophysical research communications, 493(1), 776-782 (2017-09-09)
Mitochondria are essential for cellular survival and function. In neurons, mitochondria are transported to various subcellular regions as needed. Thus, defects in the axonal transport of mitochondria are related to the pathogenesis of neurodegenerative diseases, and the movement of mitochondria



Global Trade Item Number

SKUGTIN
P7405-5MG04061834394730

Questions

  1. Have you considered any alternative products that have been tested and found suitable for the primary culture of cortical neurons? This inquiry arises as the expected delivery date for the previously ordered A-003-E Poly-D-lysine has been postponed to April 2022, and the current stock is expected to be depleted soon.

    1 answer
    1. One potential replacement is item number P7405. This is the HBr form, making it more water-soluble. However, it may be necessary to conduct tests to evaluate its compatibility with your specific cell line. If needed, there are methods to prepare the solution and subsequently remove the HBr through dialysis. This polymer is of the same size (>300,000), gamma-irradiated, and has been tested for cell culture.

      Helpful?

Reviews

No rating value

Active Filters